Search
The VersaCool headless design offers you more space to work with inside and outside the bath.
The VersaCool refrigerated bath circulator was designed with a focus on creating one of the largest work areas with the smallest footprint.
Specifications | |
| Minimum temperature | -20°C |
| Maximum temperature | 150°C |
| Temperature stability | +/-0.03 1 |
| Maximum bath volume (liters) | 71 |
| Cooling capacity (W) | 425 |
| Heater capacity (kW)230V/115V | 2/1.2 |
| Maximum pressure (mbar / psi) | 1200 / 17.4 |
| Maximum flow rate (LPM / GPM) | 14.6 / 3.86 |
| Ambient temperature range | 10 to 40°C |
| Noise (dBA) | <58 |
| Voltage range (VAC / Hz) |
100-120 / 50 or 60
200-240 / 50 or 60
|
Dimensions | |
| Working area dimensions D x W x L | 15.0 x 17.3 x 29.7cm 5.9 x 6.8 x 11.7 in. |
| Overall dimensions H x W x L | 52.9 x 25.9 x 58.5cm 20.9 x 10.2 x 23.1 in. |
| Weight (kg / lbs) | 36 / 80 |
Connectivity2 | |
| Bluetooth, Ethernet, Micro USB – front, USB – front, RS232 – front / back, RS485, Remote sensor port, Smart-Vue monitor port | |
Compliance | |
| CE, RoHS, UL / CSA, WEEE | |