XPS surface analysis system from Thermo Fisher Scientific
The Thermo Scientific Nexsa G2 X-Ray Photoelectron Spectrometer (XPS) System offers fully automated, high-throughput surface analysis, delivering the data to advance research and development or to solve production problems. By integrating XPS with ion scattering spectroscopy (ISS), UV photoelectron spectroscopy (UPS), reflected electron energy loss spectroscopy (REELS), and Raman spectroscopy, it allows you to conduct true correlative analysis.
The system now includes options for sample heating and sample biasing capabilities to increase the range of possible experiments. The Nexsa G2 Surface Analysis System unlocks the potential for advances in materials science, microelectronics, nanotechnology development, and many other applications.
Nexsa G2 Surface Analysis System features
High-performance X-ray source
A new, optimized X-ray monochromator design makes it possible to select an analysis area from 10 µm to 400 µm in 5 µm steps, ensuring data is collected from the feature of interest while maximizing the signal.
Optimized electron optics
The high-efficiency electron lens, hemispherical analyzer, and detector allow for superb detectability and rapid data acquisition.
XPS sample viewing
Bring sample features into focus with the Nexsa G2 Surface Analysis System's patented optical viewing system and XPS SnapMap, which helps you pinpoint areas of interest quickly.
XPS insulator analysis
The patented dual beam flood source couples low-energy ion beams with very low energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need, in most cases, for charge referencing. 
XPS depth profiling
Go beyond the surface with the EX06 ion source. Automated source optimization and gas handling ensure excellent performance and experimental reproducibility.
Optional XPS sample holders
Expand your possibilities with available specialist sample holders for angle-resolved XPS, sample bias measurements, or inert transfer from a glove box.
Avantage Software
Instrument control, data processing, and reporting are all controlled from Thermo Scientific Avantage Software. Avantage Software includes Knowledge View, a comprehensive source of tutorials and reference material that guide users through analysis.
Correlative XPS imaging and surface analysis workflow
Correlate XPS data with SEM images through the correlative imaging and surface analysis  workflow using Thermo Scientific Maps Software.
Nexsa G2 Surface Analysis System specifications
Analyzer type
  • 180°, double-focusing, hemispherical analyzer with 128-channel detector
X-ray source type
  • Monochromated, micro-focused, low-power Al K-Alpha X-ray source
X-ray spot size
  • 10–400 µm (adjustable in 5 µm steps)
Depth profiling
  • EX06 monatomic ion source or MAGCIS dual-mode ion source
Maximum sample area
  • 60 x 60 mm
Maximum sample thickness
  • 20 mm
Vacuum system
  • Two turbo molecular pumps, with automated titanium sublimation pump and backing pump 
Optional accessories
  • UPS, ISS, REELS, Raman spectrometer, MAGCIS, sample tilt module, NX sample heating module, sample bias module, vacuum transfer module, adaptor for glove box integration
XPS analysis resources
Nexsa G2 Surface Analysis System webinar
On-demand webinar: Thermo Scientific Nexsa G2 Surface Analysis System demo
Learn how the Nexsa G2 XPS System can be used to investigate a wide range of materials using XPS, UPS, ISS, REELS, Raman spectroscopy, and more.
XPS analysis videos
XPS analysis application notes
Nexsa G2 Surface Analysis System documentation
For Research Use Only. Not for use in diagnostic procedures.