Search
Thermo Scientific process mass spectrometers are engineered to meet a number of challenging process applications in the petrochemical, iron and steel, clean energy, and biotechnology industries. Use this comparison chart to see which process mass spectrometer is right for you.
| Model | ||||
| Overview | Maximize product yield and increase profits. This online mass spectrometer & process gas analyzer is engineered to meet a number of challenging process applications in the petrochemical, iron and steel, clean energy and biotechnology industries. | Help ensure plant safety by enabling leak detection and correction before any toxic hazard is created. This next-generation environmental mass spectrometer is engineered to help plants monitor fugitive emissions of toxic organic vapors, protecting workers and surrounding environment from hazardous exposure. | Achieve high scanning speeds and reproducible measurements. Specifically designed for process development laboratories, this real-time process gas analyzer is a highly precise, multi-stream magnetic sector analyzer. | Perform routine continuous measurement of contaminants including water and oxygen to single-digit ppt levels. Recommended for UHP Nitrogen, UHP Argon, UHP Helium, UHP Hydrogen. |
| Measurement technology | High precision scanning magnetic sector mass spectrometer | High precision scanning magnetic sector mass spectrometer | High precision scanning magnetic sector mass spectrometer | APIMS δQ: 1x Triple filter quadrupole; APIMS Quattro: 4x Triple filter quadrupole |
Our process mass spectrometer instruments are versatile gas analyzers designed to support multiple applications. Explore our applications page.
For Research Use Only. Not for use in diagnostic procedures.