Search
We have deep expertise and many years of experience formulating Applied Biosystems TaqMan master mixes for sensitive and specific detection of microbial and pathogen RNA and DNA targets for applications in research and bio-pharmaceutical discovery. Real-time PCR detection of microbial and pathogen targets requires exceptional sensitivity and accuracy, and we have master mixes that are designed specifically for this purpose.
|
|
|
Key application |
Microbial and pathogen detection (RNA targets) |
Microbial and pathogen detection (DNA targets) |
Target |
RNA |
DNA |
Concentration |
4X |
2X |
Multiplexing and passive reference dye |
Available in 2 versions: With ROX: up to 3 targets No ROX: 3 or more targets |
Available in 2 versions: With ROX: up to 3 targets Without ROX: 3 or more targets |
Cycling mode/run time |
Fast: ~60 min Standard: ~90 min |
Fast: ~50 min Standard: ~120 min |
Carryover contamination control |
||
Reverse transcriptase |
Thermostable MMLV RT included in master mix |
N/A |
Assay recommendations |
||
|
TaqMan master mixes are For Research Use Only. Not for use in diagnostic procedures.
TaqPath master mixes are general purpose reagents labeled For Laboratory Use.