Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Proteins | |||
| Insulin | 5808.0 | 1000.0 | 0.1721763 |
| Transferrin | 80000.0 | 550.0 | 0.006875 |
| Trace Elements | |||
| Sodium Selenite | 173.0 | 0.67 | 0.0038728325 |
51500 - Insulin, Transferrin, Selenium, Ethanolamine Solution (ITS-X)
Cat No:
51500056
51300 - Insulin, Transferrin, Selenium, Sodium Pyruvate Solution (ITS-A)
Cat No:
51300044