Delivering reliable and consistent results to your desktop—automatically 
Thermo Scientific Phenom Pure, Pro, and ProX G6 Desktop SEMs are economical and easy-to-use solutions that automate most of the work for you. Each model was built with productivity in mind and features a high-brightness cerium hexaboride (CeB6) source that delivers great image quality and long-term reliability.
These SEMs give reliable, consistent results faster. Even novice users can obtain high-resolution images within minutes—helping you and your team be more productive.
  • Phenom Pure G6 Desktop SEM: The most basic and economical option, suited to educational purposes and basic R&D.
  • Phenom Pro G6 Desktop SEM: An advanced imaging system for a wide range of applications.
  • Phenom ProX G6 Desktop SEM: The most advanced model, combining all the features of the Phenom Pro G6 Desktop SEM with integrated EDS for comprehensive X-ray elemental analysis. 
Phenom G6 Desktop SEM features
Phenom Pure G6 Desktop SEM
Entry-level SEM with high-brightness CeB6 source, suited to educational and basic R&D tasks. User friendly and cost effective.
Phenom Pro G6 Desktop SEM
Advanced SEM with high-quality imaging, multiple acceleration voltages, and quick operation. Intuitive design and low maintenance.
Phenom ProX G6 Desktop SEM
High-performance SEM with integrated EDS for detailed X-ray analysis. Offers comprehensive imaging and advanced analytical capabilities.
Built for your biggest needs
Whether you’re managing a lab or a quality department, Phenom G6 Desktop SEMs can help you balance budgets and quality requirements.
Phenom G6 Desktop SEM resources
Would you like to know more about Phenom G6 Desktop SEMs? Dive into a wealth of resources, including eBooks, datasheets, and more. 
Phenom G6 Desktop SEM image gallery
See for yourself what these great-value desktop SEMs can deliver for you and your team. 
For Research Use Only. Not for use in diagnostic procedures.