Dionex ICS-6000 HPIC system
You have the freedom to explore in ion chromatography
The ability to develop and run different methods for a single sample or for different samples is increasingly important for analytical laboratories. A highly flexible ion chromatography (IC) system provides you with the freedom to develop, explore, and run different methods simultaneously.
Which Dionex ICS-6000 HPIC system configuration is best for you?
Dionex ICS-6000 Standard Bore and Microbore HPIC Systems |
Dionex ICS-6000 Capillary HPIC Systems |
Dionex ICS-6000 Hybrid HPIC System |
Major benefit |
|
---|---|---|---|---|
Request quote | Request quote | Request quote | ||
Column configurations | Two standard bore 4 mm, two microbore 2 mm, or one of each column | Two capillary 0.4 mm columns | One standard bore and one capillary, or one microbore and one capillary column | Can run two different analyses simultaneously |
Column compartment | Dionex ICS-6000 DC Detector/Chromatography Compartment | Dionex ICS-6000 Capillary IC Cube | Dionex ICS-6000 Capillary IC Cube | Allows easy column installation and connections |
Column choices |
|
Offers broad variety of quality columns | ||
Detectors (standard) | Can be used in most common IC applications | |||
Detectors (optional) | Allows addition of detector for special applications | |||
6000 psi pumps | Can perform high pressure IC or, with dual pumps, 2D-IC | |||
Eluent generator | Capillary EG lasts 18 months; uses 5.25L water/day | |||
Autosampler compatibility | Offers a variety of cost and application options | |||
CDS software | Provides, power, simplicity and convenience |
A collection of IC applications for inorganics, cations, disinfection byproducts and contaminants in drinking water.
Resources