Search
Thermo Scientific products for ion chromatography are designed to keep your research moving forward seamlessly. Our innovative ion exchange chromatography column chemistries, automated eluent generation products, autosamplers, detectors, user-friendly chromatography data system, and other products help you solve your chromatographic analysis challenges.
Our wide selection of ion exchange column chemistries is designed to address a broad array of application challenges. Our Thermo Scientific Dionex IonPac columns are made for hydroxide, carbonate, and methanesulfonic acid (MSA) eluents, while our BioIC columns — the Thermo Scientific Dionex AminoPac and Thermo Scientific Dionex CarboPac columns — enable direct analysis of amino acids and carbohydrates by ion chromatography. This diverse portfolio of columns ensures that you will be able to find the optimal column for your IC analysis.
Columns used for ion exchange chromatography:
Easily meet today’s ion chromatography challenges and ensure the success of your analyses using our next-generation IC suppressors and eluent generation solutions. Our next-generation suppressors enable the highest sensitivity and lowest detection limits for determination of inorganic analytes while our eluent generation solutions ensure you get reliable, reproducible results every time.
Save time and reduce errors by automating your sample handling tasks using one of our Thermo Scientific Dionex ion chromatography autosamplers. We offer a range of products including high-precision, low-cost, metal-free autosamplers that enable low-to-high sample injection volumes and optional temperature control.
For maximum flexibility, our Thermo Scientific Dionex Integrion HPIC system and Thermo Scientific Dionex ICS-6000 HPIC system are available with internal suppressed conductivity or electrochemical detectors (DC or integrated amperometry). Alternatively, these systems can be connected to UV-Vis absorbance (variable wavelength or photodiode array), inductively coupled mass spectrometry (ICP-MS) or mass spectrometry (MS) detection. Both the Thermo Scientific Dionex Inuvion Core IC and Thermo Scientific Dionex Inuvion IC system come with an integrated, thermally controlled conductivity detector as standard.
Create easy, consistent, low-dead-volume connections that maximize the performance of your Thermo Scientific Dionex ion chromatography system using Thermo Scientific Dionex IC PEEK Viper fittings. These fittings ensure that the ferrule is correctly positioned for increased efficiency and sharp chromatographic peaks and are available in convenient kits tailored specifically to your IC system.
A chromatography data system (CDS) is vital for efficient, reliable, and compliant handling of your chromatographic and MS data and management of the entire analytical process. Thermo Scientific Chromeleon 7 CDS is designed to be easy to use and to address the needs of the laboratory as well as the needs of IT. It is fast and intuitive, minimizing training time and driving increased lab efficiency. It also provides a resilient client-server architecture with industry-leading network failure protection to ensure data security and deliver uninterrupted operation.
Chromeleon CDS unlocks the full features and capabilities of your Thermo Scientific Dionex ion chromatography system, ensuring your instrument operates to its full potential.
Ensure the best chromatographic results by using quality parts and accessories designed specifically for your Thermo Scientific Dionex ion chromatography system and its related modules. We offer parts and accessories for:
To help simplify your ion chromatography analyses, we have developed all the high-purity reagents and standards you need. Our complete line of Thermo Scientific Dionex standards, reagents, and chemicals can help minimize time-consuming reagents and standards validation and keep your system operating at peak performance. Our selection of IC standards and reagents includes:
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.