Human antibody cross-reactivity
SpecificityCloneRhesusCynomolgus monkeyBaboonChimpanzeeMarmosetPigCanine
Bcl-2Bcl-2/100++     
CD1aHI149---    
CD1aSK9+ +
CD2RPA-2.10++++++ 
CD3HIT3a---+ - 
CD3OKT3-  +   
CD3SK7 -
 +
CD3UCHT1---+ - 
CD4OKT4++-+   
CD4RPA-T4---+ - 
CD4SK3++     
CD5L17F12--     
CD5UCHT2---++- 
CD74H9--     
CD8HIT8a+--+ - 
CD8OKT8+++ -  
CD8RPA-T8++++-- 
CD8SK1++  -  
CD11aHI111++++ - +
CD11bICRF44+++++- 
CD11bM1/70++     
CD11c3.9++
+
+
-  
CD13WM15---+ - 
CD1461D3+    - 
CD15HI98---+ - 
CD163G8+++++  
CD186.7++++   
CD19HIB19--++-- 
CD19SJ25C1- -
CD202H7++++-- 
CD20L26++     
CD21HB5    +  
CD22S-HCL-1++     
CD25BC96+(weak)++ -  
CD25CD25-4E3+   - -
CD26M-A261 - +   
CD27O323+++    
CD28CD28.2++++ - 
CD31WM-59+++- - 
CD33HIM3-4---+  -
CD33P67.6--
    -
CD33WM53---+ - 
CD35E11+++++  
CD36NL07   +   
CD38HB7--  -
CD38HIT2---+ - 
CD39eBioA1 (A1)+    - 
CD405C3++++   
CD41aHIP8++++ + 
CD41bHIP2++++   
CD42bHIP1   +   
CD44IM7++++
 + 
CD452D1-      
CD45HI30-- + - 
CD45RRA3-6B2  -+   
CD45RAGRT22+-     
CD45RAHI100---+   
CD45ROUCHL1---+ -+
CD49d9F10+++++-+
CD49feBioGoH3 (GoH3)++++  +
CD51/6123C6---- - 
CD54HA58---+ - 
CD54RR1/1+      
CD56TULY56+      
CD57TBO1- -    
CD61VI-PL2++++  +
CD62LDREG-56 (DREG56)---+ - 
CD62LSK11++++ + 
CD62PAK-4---+   
CD6410.1++++ - 
CD68eBioY1/82A (Y1/82A) +     
CD69FN50++++ - 
CD73AD2+  +   
CD74LN2+-+    
CD79aHM47+
+
+
+
  +
CD79bCD3-1 - -   
CD802D10+      
CD83HB15e++++ - 
CD842G7---+   
CD85d42D1---    
CD85jHP-F1---    
CD85kZM4.1-      
CD86IT2.2++++   
CD87VIM5++++   
CD90eBio5E10 (5E10)+++  + 
CD94DX22---    
CD95DX2++++ -+
CD97VIM3b---+   
CD101BB27     - 
CD102CBRIC2/2+      
CD103Ber-ACT8 +     
CD103B-Ly7+      
CD105SN6+      
CD107aeBioH4A3+    - 
CD107beBioH4B4 (H4B4)+      
CD1164H1---    
CD117104D2++     
CD117YB5.B8      +
CD119 (IFNgR1 alpha chain)GIR-208 (GIR 208)+      
CD1236H6+      
CD127eBioRDR5+   +  
CD132TUGh4---+   
CD134ACT35 (ACT-35)--     
CD1374B4 (4B4-1)++++   
CD146P1H12      +
CD150A12 (7D4)
+++-
CD15214D3+      
CD154 (CD40 Ligand)
24-31+
+     
CD160BY55+--    
CD161HP-3G10+      
CD163eBioGHI/61 +   +
CD1663A6   +   
CD177MEM-166+      
CD180MHR73-11+++    
CD184 (Fusin)12G5++++   
CD185 (CXCR5)MU5UBEE+      
CD196 (CCR6)R6H1+      
CD197 (CCR7)
3D12+      
CD205MG38+      
CD20619.2++++   
CD243UIC2-      
CD244C1.7---    
CD275 (ICOS-L)MIH12-      
CD279 (PD-1)eBioJ105 (J105)+      
CD279 (PD-1)MIH4-      
CD281 (TLR1)GD2.F4+      
CD282 (TLR2)TL2.1+      
CD357 (AITR/GITR)eBioAITR +     
Foxp3236A/E7 +     
Foxp3PCH101++++-  
GARPG14D9       
GM-CSFBVD21C11+      
HLA-ABCW6/32+++    
HLA-DRL243++  + +
HLA-DRLN3 +     
IFN gamma4S.B3+
+
+
+
IFN gammaMD-1++++   
IL-1a364/3B3-14+      
IL-2MQ1-17H12++++   
IL-4MP4-25D2+      
IL-48D4-8++++   
IL-10JES3-9D7+++    
IL-12 (P40/70)C8.6+      
IL-13PVM13-1+      
IL-17AeBio64CAP17++
IL-17AeBio64DEC17+     +
IL-22IL22JOP+      
Integrin beta7FIB504++
++   
Ki-6720Raj1      +
Lambda light chainJDC-12 +     
MCP-1 (CCL2)5D3-F7++   + 
T-beteBio4B10+      
TCR alpha/betaIP26---    
TCR gamma/deltaB1.1++++   
TLR1 (CD281)
GD2.F4+      
TLR2 (CD282)
TL2.1+
     +
TLR3TLR3.7+     +
TLR4HTA125+ +   +
TNF alphaMab11+++    
TNF beta359-81-11+      
For Research Use Only. Not for use in diagnostic procedures.