CellInsight CX7 LZR High-Content Screening (HCS) Platform
The Thermo Scientific CellInsight CX7 LZR High-Content Screening (HCS) Platform is an integrated system for advanced performance in screening and analysis.
Request a demoRequest a quoteRequest more information
Demonstrating the CellInsight CX7 LZR
Learn more about high-content cell analysis software
Learn more about high-content screening applications
Image of a scientist at a high-content screening instrument
Even more advanced performance through powerful laser illumination light source
CellInsight CX7 LZR HCS platform is fast, laser-based, automated cellular imaging and analysis platform for quantitative microscopy and phenotypic screening. It’s designed to provide you with the sensitivity and speed that’s needed for emerging assays.
CellInsight CX7 LZR system highlights
Illumination 7-channel, laser-based illumination
Camera Photometrics High-Resolution Fluorescent Camera
Widefield 7-channel fluorescent imaging
Brightfield 5-channel brightfield imaging
Confocal 7-channel confocal imaging
Objectives 3-position objective (range from 2x–40x, low and high NA)
Focus Software and laser-based autofocus for consistent scan times
Automation Configured for fully automated plate handling and scanning
Software HCS Studio software for integrated data collection and analysis
HeLa cells imaged using the CellInsight CX7 platform
HeLa cell image acquired using the Thermo Scientific CellInsight CX7 platform and stained with Invitrogen HCS NuclearMask Blue (blue), Invitrogen MitoTracker Orange (green), and Invitrogen Alexa Fluor 647 Phalloidin (red).
A549 cells imaged using the CellInsight CX7 platform
A549 cell image acquired using the Thermo Scientific CellInsight CX7 platform and stained with Invitrogen HCS CellMask Blue (blue), Invitrogen Alexa Fluor 488 Phalloidin (green), and Invitrogen Alexa Fluor 750 goat anti–mouse IgG secondary antibody (pink).
Three-color brightfield image of a fixed tissue slice imaged with the CellInsight CX7 platform.
Three-color brightfield image of fixed human breast cancer tissue stained with KI67-DAB and counterstained with hematoxylin and eosin and imaged with the Thermo Scientific CellInsight CX7 platform.
Reagents optimized for high-content screening
Drawing on decades of experience in fluorescence imaging, Invitrogen HCS products are developed using Thermo Scientific High-Content Platforms with special considerations for the high-throughput workflow and automated imaging:
  • Cell and nuclear masks for automated demarcation
  • Robust functional probes for cell health interrogation
  • Flexible assay workflow for automated processing
  • Broad fluorophore choice for easy multiplexing
  • Validated on multiple imaging platforms
Select reagents for use with the CellInsight CX7 LZR
Other cell structure and segmentation tools
Clear identification of cell and nuclear boundaries in automated imaging requires fiducial markers that will label cellular structures robustly and reproducibly. Cell masks are designed specifically for automated imaging. Molecular Probes cell structure labels are widely used in microscope imaging and HCS.
See other reagents for cell structure
Other cell health and cytotoxicity assays
We offer a diverse selection of imaging and microscopy assays for the analysis of cell viability, proliferation, apoptosis, cell cycle and DNA repair, migration, adhesion and other cellular functions. Many of these assays have been validated specifically for HCS platforms.
Browse cell health and cytotoxicity assays by application:
Microplates and slides
Compatible with SBS standard microplates (6–1,536 well) and slides
Automation
Orbitor RS 2 Microplate Mover
Increase throughput, storage capacity and operational flexibility with the Thermo Scientific Orbitor RS 2 Microplate Mover—industrial-sized performance within a compact, benchtop mover.
The Orbitor RS 2 is a high-speed microplate mover offering proven, reliable performance with totally flexible plate handling. Extensive vertical reach allows multiple stacked or high density instruments to be loaded in a small footprint, and a bi-directional telescoping arm provides superior reach, improved user safety and unlimited base rotations within a 360° workspace.
Orbitor RS 2 Microplate Mover
Camera Photometrics High-Resolution Fluorescent Camera:
  • High sensitivity, thermoelectrically cooled to 0°C
  • 14-bit data acquisition and processing
  • Field of view (FOV): 2,208 x 2,208 pixel array
  • 4.54 μm pixel size
  • 75% peak quantum efficiency (QE)
  • Multiple pixel binning options for flexible analysis options
Data management Compatible with Thermo Scientific Store Image and Database Management Software
Dimensions 20 in. x 32 in. x 18 in. (50.8 cm x 81.3 cm x 45.7 cm)
Image compatibility JPG, BMP, GIF, PNG, TIF, C01, DIB
Light source
High-output, laser-based illumination. Configured for imaging in the UV range through near-IR range with the provided filter sets.
Objectives
Standard (Olympus™ objectives):
  • 10x (0.30 NA) Plan NeoFluar™ system
  • 20x (0.4 NA) Achroplan™ system
  • 20x (0.7 NA) Olympus objectives
  • 40x (0.6 NA) Olympus objectives
Optional (Olympus objectives):
  • 2x (0.08 NA)
  • 4x (0.16 NA)
PC Windows 7 Professional 64-Bit PC with 32GB (4x8GB) RAM, Intel® Xeon® Processor 3.7GHz Turbo, RAID I Controller with 1.8TB Hard Drive and 256 Boot Drive. Includes Dell 24" High Resolution Widescreen Monitor or equivalent, keyboard and mouse.
Software Thermo Scientific HCS Studio 4.0 Cell Analysis Software
Wattage 300 W
Weight 150 lb (68 kg)
Reagents optimized for high-content screening
Drawing on decades of experience in fluorescence imaging, Invitrogen HCS products are developed using Thermo Scientific High-Content Platforms with special considerations for the high-throughput workflow and automated imaging:
  • Cell and nuclear masks for automated demarcation
  • Robust functional probes for cell health interrogation
  • Flexible assay workflow for automated processing
  • Broad fluorophore choice for easy multiplexing
  • Validated on multiple imaging platforms
Select reagents for use with the CellInsight CX7 LZR
Other cell structure and segmentation tools
Clear identification of cell and nuclear boundaries in automated imaging requires fiducial markers that will label cellular structures robustly and reproducibly. Cell masks are designed specifically for automated imaging. Molecular Probes cell structure labels are widely used in microscope imaging and HCS.
See other reagents for cell structure
Other cell health and cytotoxicity assays
We offer a diverse selection of imaging and microscopy assays for the analysis of cell viability, proliferation, apoptosis, cell cycle and DNA repair, migration, adhesion and other cellular functions. Many of these assays have been validated specifically for HCS platforms.
Browse cell health and cytotoxicity assays by application:
Microplates and slides
Compatible with SBS standard microplates (6–1,536 well) and slides
Automation
Orbitor RS 2 Microplate Mover
Increase throughput, storage capacity and operational flexibility with the Thermo Scientific Orbitor RS 2 Microplate Mover—industrial-sized performance within a compact, benchtop mover.
The Orbitor RS 2 is a high-speed microplate mover offering proven, reliable performance with totally flexible plate handling. Extensive vertical reach allows multiple stacked or high density instruments to be loaded in a small footprint, and a bi-directional telescoping arm provides superior reach, improved user safety and unlimited base rotations within a 360° workspace.
Orbitor RS 2 Microplate Mover
Camera Photometrics High-Resolution Fluorescent Camera:
  • High sensitivity, thermoelectrically cooled to 0°C
  • 14-bit data acquisition and processing
  • Field of view (FOV): 2,208 x 2,208 pixel array
  • 4.54 μm pixel size
  • 75% peak quantum efficiency (QE)
  • Multiple pixel binning options for flexible analysis options
Data management Compatible with Thermo Scientific Store Image and Database Management Software
Dimensions 20 in. x 32 in. x 18 in. (50.8 cm x 81.3 cm x 45.7 cm)
Image compatibility JPG, BMP, GIF, PNG, TIF, C01, DIB
Light source
High-output, laser-based illumination. Configured for imaging in the UV range through near-IR range with the provided filter sets.
Objectives
Standard (Olympus™ objectives):
  • 10x (0.30 NA) Plan NeoFluar™ system
  • 20x (0.4 NA) Achroplan™ system
  • 20x (0.7 NA) Olympus objectives
  • 40x (0.6 NA) Olympus objectives
Optional (Olympus objectives):
  • 2x (0.08 NA)
  • 4x (0.16 NA)
PC Windows 7 Professional 64-Bit PC with 32GB (4x8GB) RAM, Intel® Xeon® Processor 3.7GHz Turbo, RAID I Controller with 1.8TB Hard Drive and 256 Boot Drive. Includes Dell 24" High Resolution Widescreen Monitor or equivalent, keyboard and mouse.
Software Thermo Scientific HCS Studio 4.0 Cell Analysis Software
Wattage 300 W
Weight 150 lb (68 kg)
Learn more about 2D/3D visualization and analysis (2D-5D)
Amira Software for Life & Biomedical Sciences Brochure cover
With incredible speed and flexibility, Amira Software supports advanced 2D–5D bioimaging workflows in research areas ranging from structural and cellular biology to tissue imaging, neuroscience, preclinical imaging and bioengineering.
Download the Amira software guide
Learn more about Amira software