eFluor organic dyes spectra
Antibody conjugates offering superior optical performance
Invitrogen eBioscience eFluor organic dyes were developed specifically for labeling and detection of biomolecules. Each eFluor dye is engineered for superior optical performance in a variety of applications.
Reliable performance in multiple applications
eFluor organic dyes are traditional fluorescent dyes for use in multiple applications, including multicolor flow cytometry and immunofluorescence microscopy. To make fluorophore selection easier, all eFluor organic dyes are named after their emission wavelength and they are fully compatible with most conventional dyes.
Dual parameter scatter plot
eFluor organic dyes
Dye eFluor 450 eFluor 506
Fluorophore type Organic dye Organic dye
Ex (nm) 401 419
Em (nm) 421 506
Application Flow cytometry Flow cytometry
Conjugates available eFluor 450 primary antibody conjugates
eFluor 450 secondary antibody conjugates
eFluor 506 primary antibody conjugates
Recommended bandpass filter 450/50 510/50
Uses Comparable to Invitrogen Pacific Blue dye Use in place of AmCyan or Horizon V500 dye
Dye eFluor 520 eFluor 570 PE-eFluor 610 PerCP-eFluor 710
Fluorophore type
Organic dye
Organic dye
Organic dye (tandem), resonance energy transfer from PE molecule to eFluor 610 dye
Organic dye (tandem), resonance energy transfer from PerCP molecule to eFluor 710 dye
Ex (nm)
475
546
496, 565
488
Em (nm)
535
585
607
710
Application
ICC, IHC
ICC, IHC
FC
FC
Conjugates available
eFluor 570 primary antibody conjugates
eFluor 570 secondary antibody conjugates
PE-eFluor 610  primary antibody conjugates
PE-eFluor 610  secondary antibody conjugates
PerCP-eFluor 710 primary antibody conjugates
PerCP-eFluor 710 secondary antibody conjugates
Recommended bandpass filter
535/45
585/40
610/20
695/40
Uses
Alternative to Invitrogen Alexa Fluor 488 dye
Alternative to Invitrogen Alexa Fluor 555 dye
Alternative to Invitrogen PE-Texas Red dye
(Note: PE-eFluor 610 is brighter and requires reduced compensation from far-red channels compared to PE-Texas Red dye)
Alternative to PE-Cyanine 5.5 dye or PerCP-Cyanine 5.5
Dye eFluor 570 eFluor 615
Fluorophore type Organic dye Organic dye
Ex (nm) 546 560
Em (nm) 585 645
Application ICC, IHC ICC, IHC
Conjugates available
eFluor 570 primary antibody conjugates
eFluor 570 secondary antibody conjugates
eFluor 615 primary antibody conjugates
eFluor 615 secondary antibody conjugates
Recommended bandpass filter 585/40 645/75
Uses Alternative to Invitrogen Alexa Fluor 555 dye Alternative to Invitrogen Alexa Fluor 647 dye
Dye eFluor 660 APC-eFluor 780
Fluorophore type Organic dye Organic dye (tandem), resonance energy transfer from APC molecule to eFluor 780 dye
Ex (nm) 633 645
Em (nm) 668 767
Application Flow cytometry, ICC, IHC Flow cytometry
Conjugates available
eFluor 660 primary antibody conjugates
eFluor 660 secondary antibody conjugates
APC-eFluor 780 primary antibody conjugates
APC-eFluor 780 secondary antibody conjugates
Recommended bandpass filter 660/20 780/60
Uses Alternative to APC or Invitrogen Alexa Fluor 647 dye Comparable to APC-Cyanine 7 and Invitrogen APC-Alexa Fluor 750 dye
Resources
Custom Services
Contact our eBioscience custom services group for antibody conjugation, antibody cocktails, immunoassay development, bulk reagents, and testing services.
Support
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.