Thermo Scientific TSX series refrigerators are high performance units designed with features that support sample protection and sustainability for storing media, reagents, chemotherapy and pharmaceutical compounds, and other medical- and laboratory-grade products requiring a storage environment of 3° to 7°C. The series’ V-drive technology, coupled with Phononic™ heat pumps for our undercounter units, is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
- V-drive offers quiet, reliable performance
- Energy Star certified
- Industry-best 10 year compressor warranty
Browse our complete portfolio or contact one of our experts to help you make a selection.
Quick links
Choose a compact high performance lab refrigerator
TSX Glass Door Undercounter Refrigerator |
TSX Solid Door Undercounter Refrigerator |
|
![]() |
![]() |
|
The TSX Series Undercounter Refrigerators, powered by Phononic solid state refrigeration technology, have been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required, in support of breastmilk and medication storage, analyzer kits and reagents, and even cell culture media, featuring an independent/reduntant cooling system, data-logging, and access ports to provide the ultimate compact, sample storage solution. Glass or solid door moldels support requirements of research for monitoring the most rigorous and demanding storage needs, all while using less energy and putting almost no heat into the work environment. |
||
Request a quote |
||
Capacity | 5.5 cu. ft. (156L) | 5.5 cu. ft. (156L) |
Temperature Range | 3° to 7°C | 3° to 7°C |
Doors | 1 | 1 |
Shelves | 3 | 3 |
Interior (DxWxH) | 20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4cm) | 20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4cm) |
Exterior (DxWxH) | 26 x 23.6 x 31.8 in. (66 x 60 x 81cm) | 26 x 23.6 x 31.8 in. (66 x 60 x 81cm) |
Shipping Weight | 155 lb. (71kg) | 155 lb. (71kg) |
Software downloads for TSX Undercounter Refrigerators
Instructions for User Interface (UI) Firmware Updates
TSX505 Software Update v1.1.03
Download (zip file)
TSX505 Software Update v1.2.16
Download (zip file)
TSX505 Software Update v1.3.00
Download (zip file)
Looking for refrigerators specifically designed for vaccine storage? Visit our Vaccine Storage Solutions page.
Choose an upright high performance lab refrigerator
Looking for refrigerators for medical-related applications?
Visit our Upright Medical Refrigerators & Freezers page to browse and learn more about our high performance refrigerators designed with special temperature control, access and organizational features.
Single door high performance lab refrigerators
Glass door
11.5 cu. ft. Glass Door Refrigerator |
23 cu. ft. Glass Door Refrigerator |
29.2 cu.ft. Glass Door Refrigerator |
|
![]() |
![]() |
![]() |
|
V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
|
|||
Request a quote |
|||
Capacity | 11.5 cu. ft. (326L) | 23 cu. ft. (650L) | 29.2 cu. ft. (827L) |
Temperature Range | 2° to 8°C | 2° to 8°C | 2° to 8°C |
Doors | 1 | 1 | 1 |
Shelves | 4 | 4 | 4 |
Interior (DxWxH) | 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm) | 28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm) | 28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4cm) | 37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6cm) | 37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4cm) |
Shipping Weight | 339 lb. (154kg) | 450 lb. (204kg) | 458 lb. (208kg) |
11.5 cu. ft. (326L) |
11.5 cu. ft. (326L) |
Solid door
11.5 cu. ft. Solid Door Refrigerator |
23 cu. ft. Solid Door Refrigerator |
29.2 cu. ft. Solid Door Refrigerator |
|
![]() |
![]() |
![]() |
|
Request a quote |
|||
Capacity | 11.5 cu. ft. (326L) | 23 cu. ft. (650L) | 29.2 cu. ft. (827L) |
Temperature Range | 2° to 8°C | 2° to 8°C | 2° to 8°C |
Doors | 1 | 1 | 1 |
Shelves | 4 | 4 | 4 |
Interior (DxWxH) | 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm) | 28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm) | 28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4cm) | 37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6cm) | 37.8 x 34.0 x 78.5 in. (96.0 x 86.4 x 199.4cm) |
Shipping Weight | 340 lb. (154kg) | 426 lb. (193kg) | 459 lb. (208kg) |
11.5 cu. ft. (326L) |
11.5 cu. ft. (326L) |
11.5 cu. ft. Glass Door Refrigerator |
11.5 cu. ft. Solid Door Refrigerator |
29.2 cu. ft. Glass Door Refrigerator |
29.2 cu. ft. Solid Door Refrigerator |
|
![]() |
![]() |
![]() |
![]() |
|
Store and retrieve pharmacy products with ease, utilizing Thermo Scientific™ Forma™ Refrigerators. Developed for stable temperature control over a range of 1° to 8°C, these refrigerators feature microprocessor control, positive airflow systems, industrial-quality cabinet construction, and extra-strength refrigeration compressors.
|
||||
Request a quote |
||||
Capacity | 11.5 cu. ft. (326L) | 11.5 cu. ft. (326L) | 29.2 cu. ft. (827L) | 29.2 cu. ft. (827L) |
Temperature Range | 1° to 8°C | 1° to 8°C | 1° to 8°C | 1° to 8°C |
Doors | 1 | 1 | 1 | 1 |
Shelves | 4 |
4 | 4 | 4 |
Interior (DxWxH) | 21 x 20 x 52.4 in. (55.2 x 50.8 x 133.1cm) | 21 x 20 x 52.4 in. (55.2 x 50.8 x 133.1cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm) | 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm) |
Shipping Weight | 220 lb. (100kg) | 280 lb. (127kg) | 435 lb. (197kg) | 435 lb. (197kg) |
11.5 cu. ft. Glass Door Refrigerator |
11.5 cu. ft. Solid Door Refrigerator |
29.2 cu. ft. Glass Door Refrigerator |
29.2 cu. ft. Glass Door Refrigerator |
|
![]() |
![]() |
![]() |
![]() |
|
Store and retrieve pharmacy products with ease, utilizing Thermo Scientific™ Revco™ Refrigerators. Developed for stable temperature control over a range of 1° to 8°C, these refrigerators feature microprocessor control, positive airflow systems, industrial-quality cabinet construction, and extra-strength refrigeration compressors.
|
||||
Request a quote |
||||
Capacity | 11.5 cu. ft. (326L) | 11.5 cu. ft. (326L) | 29.2 cu. ft. (827L) | 29.2 cu. ft. (827L) |
Temperature Range | 1° to 8°C | 1° to 8°C | 1° to 8°C | 1° to 8°C |
Doors | 1 | 1 | 1 | 1 |
Shelves | 4 | 4 | 4 | 4 |
Interior (DxWxH) | 21.75 x 20 x 52.4 in. (55.3 x 50.8 x 133.1cm) | 21 x 20 x 52.4 in. (55.3 x 50.8 x 133.1cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) | 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm) |
Shipping Weight | 280 lb. (127kg) | 280 lb. (127kg) | 435 lb. (197kg) | 425 lb. (193kg) |
11.5 cu. ft. Glass Door Refrigerator |
11.5 cu. ft. Solid Door Refrigerator |
29.2 cu. ft. Glass Door Refrigerator |
29.2 cu. ft. Glass Door Refrigerator |
|
![]() |
![]() |
![]() |
![]() |
|
Store and retrieve pharmacy products with ease, utilizing Thermo Scientific™ Jewett™ Refrigerators. Developed for stable temperature control over a range of 1° to 8°C, these refrigerators feature microprocessor control, positive airflow systems, industrial-quality cabinet construction, and extra-strength refrigeration compressors.
|
||||
Request a quote |
||||
Capacity | 11.5 cu. ft. (326L) | 11.5 cu. ft. (326L) | 29.2 cu. ft. (827L) | 29.2 cu. ft. (827L) |
Temperature Range | 1° to 8°C | 1° to 8°C | 1° to 8°C | 1° to 8°C |
Doors | 1 | 1 | 1 | 1 |
Shelves | 4 |
4 | 4 | 4 |
Interior (DxWxH) | 20 x 21 x 52.4 in. (50.8 x 53.3 x 133.1cm) | 20 x 21 x 52.4 in. (50.8 x 53.3 x 133.1cm) | 30 x 29 x 58 in. (76.2 x 73.7 x 147.3cm) | 30 x 29 x 58 in. (76.2 x 73.7 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 30 x 73.6 in. (74.3 x 76.2 x 186.9cm) | 29.3 x 30 x 73.6 in. (74.3 x 76.2 x 186.9cm) | 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm) | 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm) |
Shipping Weight | 220 lb. (127kg) | 280 lb. (127kg) | 435 lb. (197kg) | 435 lb. (197kg) |
Double door high performance lab refrigerators
51 cu. ft. Glass Door Refrigerator |
51 cu. ft. Solid Door Refrigerator |
|
![]() |
![]() |
|
V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
|
V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
|
|
Request a quote |
||
Capacity | 51 cu. ft. (1447L) | 51 cu. ft. (1447L) |
Temperature Range | 2° to 8°C | 2° to 8°C |
Doors | 2 | 2 |
Shelves | 8 | 8 |
Interior (DxWxH) | 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm) | 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm) |
Exterior (DxWxH) | 37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4cm) | 37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4cm) |
Shipping Weight | 669 lb. (303kg) | 621 lb. (282kg) |
Triple door high performance lab refrigerators
78.8 cu. ft. Glass Door Refrigerator |
78.8 cu. ft. Solid Door Refrigerator |
|
![]() |
![]() |
|
Store and retrieve pharmacy products with ease, utilizing Thermo Scientific™ Forma™ Refrigerators. Developed for stable temperature control over a range of 1° to 8°C, these refrigerators feature microprocessor control, positive airflow systems, industrial-quality cabinet construction, and extra-strength refrigeration compressors.
|
||
Request a quote |
||
Capacity | 78.8 cu. ft. (2231L) | 78.8 cu. ft. (2231L) |
Temperature Range | 1° to 8°C | 1° to 8°C |
Doors | 3 | 3 |
Shelves | 12 | 12 |
Interior (DxWxH) | 29 x 81 x 58 in. (73.7 x 205.7 x 147.3cm) | 29 x 81 x 58 in. (73.7 x 205.7 x 147.3cm) |
Exterior (DxWxH) | 37.2 x 85 x 79.2 in. (94.5 x 215.9 x 201.2cm) | 37.2 x 85 x 79.2 in. (94.5 x 215.9 x 201.2cm) |
Shipping Weight | 805 lb. (365kg) | 805 lb. (365kg) |
78.8 cu. ft. Glass Door Refrigerator |
78.8 cu. ft. Solid Door Refrigerator |
|
![]() |
![]() |
|
Store and retrieve pharmacy products with ease, utilizing Thermo Scientific™ Forma™ Refrigerators. Developed for stable temperature control over a range of 1° to 8°C, these refrigerators feature microprocessor control, positive airflow systems, industrial-quality cabinet construction, and extra-strength refrigeration compressors.
|
||
Request a quote |
||
Capacity | 78.8 cu. ft. (2231L) | 78.8 cu. ft. (2231L) |
Temperature Range | 1° to 8°C | 1° to 8°C |
Doors | 3 | 3 |
Shelves | 12 | 12 |
Interior (DxWxH) | 29 x 81 x 58 in. (73.7 x 205.7 x 147.3cm) | 29 x 81 x 58 in. (73.7 x 205.7 x 147.3cm) |
Exterior (DxWxH) | 37.2 x 85 x 79.2 in. (94.5 x 215.9 x 201.2cm) | 37.2 x 85 x 79.2 in. (94.5 x 215.9 x 201.2cm) |
Shipping Weight | 805 lb. (365kg) | 805 lb. (365kg) |
Which plug type do I need for my high performance refrigerator?
Standard Plug Type | Voltage | Hz | |
---|---|---|---|
![]() |
NEMA 5-15 | 115–120 | 60 |
![]() |
NEMA 5-20 | 115–120 | 60 |
![]() |
NEMA 6-15 | 208–230 | 60 |
![]() |
Continental EU |
230 | 50 |
Featured Brochure
TSX Series High-Performance Refrigerators and Freezers
Lab and clinical-grade storage that adapts to you and your environment.
Related products
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic