TSV Value Combo Refrigerator/Freezer
TSV Value Combo Refrigerator/Freezer
TSV Value Combo Refrigerator/Freezer
TSV Value Combo Refrigerator/Freezer
TSV Value Combo Refrigerator/Freezer
TSV Value Combo Refrigerator/Freezer
Thermo Scientific™

TSV Value Combo Refrigerator/Freezer

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value Refrigerator/Freezer.
Have Questions?
Catalog NumberCapacity (Metric)
TSV18CPSA510 L
Catalog number TSV18CPSA
Price (USD)
-
Capacity (Metric):
510 L
Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value Refrigerator/Freezer.
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, SNAP-compliant hydrocarbon refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto-defrost combination units
Specifications
Amperage1.1A
Cabinet MaterialCold-rolled Steel
Casters2 adjustable leveling feet
ColorWhite
Control TypeElectronic Controls
DefrostAutomatic
Dimensions (D x W x H) InteriorRefrigerator: 25.5 x 25 x 40 in. (64.7 x 63.5 x 101.6 cm) Freezer: 20.5 x 22.5 x 15.75 in. (52 x 57.1 x 40 cm)
Dimensions (L x W x H) Exterior32.79 x 29.53 x 66.61 in. (83.3 x 75 x 169.2 cm)
Door Shelves3
Electrical Requirements115 V/60 Hz
Exterior FinishEnameled Steel
For Use With (Application)Indoor general storage of non-critical samples and reagents
Frequency50 Hz
Hinge SideRight
InsulationCyclopentane
InteriorABS Plastic
LightingInterior
Min. Clearance4 in. on Sides, 4 in. at Back, 11 in. on Top
Monitoring OptionsNone
RefrigerantR600a
Shelves2 Adjustable Shelves, 2 Drawers (Refrigerator); 1 Shelf (Freezer)
Temperature (Setpoint)Adjustable
TypeCombination General Purpose Refrigerator/Freezer
Typical Lifetime10 years
Weight (English)224 lb.
Weight (Metric)101.6 kg
Capacity (English)18 cu. ft.
Capacity (Metric)510 L
Certifications/CompliancecULus
CompressorBottom-mounted
Door StyleHinged
No. of Doors2 (2 compartments)
Plug TypeNEMA 5-15 P
StyleUpright
Temperature Range1°C to 12°C
Voltage115 V
Unit SizeEach