Search Thermo Fisher Scientific
Applied Biosystems custom primers and TaqMan probes are the best option for research experiments or applications that cannot be addressed with one of our predesigned TaqMan Assays. Our portfolio of fluorescent dyes and quenchers provides maximum assay design flexibility while retaining the quality, reproducibility, and gold-standard chemistry you have come to expect from TaqMan reagents.
Choose the probe that is best for your application:
Custom MGB probes—MGB probes offer the best all-around combination of sensitivity, precision, and specificity. MGB technology enables shorter probe designs, resulting in maximal sequence discrimination and target flexibility.
Custom QSY and QSY2 probes—TaqMan QSY and QSY2 probes are designed for optimal sensitivity and performance in multiplexed qPCR experiments for detection of up to six targets. Use QSY for multiplexing up to four targets with ABY, JUN, FAM, and VIC and use QSY2 for 5th and 6th target multiplexing with Cyanine 5 and Cyanine 5.5.
Custom TAMRA probes—one of the first TaqMan probes developed, these probes are ideal for maintaining consistency in projects using legacy TAMRA probe designs
Sequence detection primers—high-quality, unlabeled oligos for use with either TaqMan probes or SYBR dye for all of your real-time PCR research applications. Our primers are desalted, available in your choice of scales, and shipped as liquid or dried down.
Choose from our collection of dual-labeled TaqMan probes and unlabeled oligos for use as qPCR primers. Whether you want to simultaneously interrogate multiple targets, use your own bioinformatics to design a probe, or detect exotic targets that do not already have a predesigned assay, we offer the ideal probes to design your own assay. Our TaqMan MGB (minor groove binder), TAMRA, QSY, and QSY2 custom probes deliver outstanding performance and are manufactured using the same facilities and raw materials as the TaqMan Assays that are featured in over 40,000 publications.
The quality of your data is limited by the quality of the primers and probes you choose. Here's how Custom TaqMan probes set your assay up for success:
*4-10 Days in North America. International times may vary.
We offer 20+ million predesigned TaqMan Assays across a diverse set of real-time PCR applications, including gene expression, SNP genotyping, and copy number variation. Predesigned TaqMan Assays are ready-to-use primer and probe sets that are designed using our proprietary bioinformatics and design algorithms to detect your gene, SNP, or sequence of interest. Before ordering Custom Primers and TaqMan Probes, check to see if we already have a Predesigned Assay ready for you.
Our Specialty Manufacturing service offers even more customized solutions for those truly challenging real-time PCR experiments. From off-catalog dye offerings to specialized formulation and volume requirements to specific assay plating arrangements, our Specialty team will work with you to design your perfect assay.
Learn more about Specialty products ›
Leverage the full expertise and infrastructure of Thermo Fisher Scientific:
Learn more about our OEM and contract manufacturing services, GMP oligos, and more ›
We've partnered with Bioz, the world's most comprehensive AI search engine for scientists, to serve up scientific papers relevant to your research.
For Research Use Only. Not for use in diagnostic procedures.