Introduction
“NeuroELISA” kits are sandwich ELISAs designed for fast, accurate, and sensitive quantitation of protein biomarkers crucial for neurobiology and neuroscience research. These ready-to-use kits enable measurements from various biological samples, including serum, plasma, and cerebrospinal fluid (CSF). Manufactured to help ensure excellent quality and reproducibility, each kit meets rigorous specifications for sensitivity, dynamic range, precision, specificity, recovery, and lot-to-lot consistency. Our Invitrogen ELISA kits also offer specific detection of phosphorylation and correlate well with western blot data.
Learn more about Neurobiology ELISA Kits and Multiplex Immunoassays
Note: The ELISA assay protocol provided is representative of most ready-to-use ELISA kits for neurobiology. Protocols for individual kits may differ. Depending on the protein of interest, antibodies, buffers, or substrates being used, this general protocol may need to be optimized.
Search available Neurobiology ELISA kits  Browse available ELISA buffers
Materials
Typical Neurobiology ELISA kit components
  • Antibody-coated 96-well plate
  • Detection antibody
  • Standard
  • Standard diluent buffer
  • HRP conjugate (antibody or streptavidin)
  • HRP diluent
  • Wash buffer
  • Chromogenic substrate (TMB)
  • Stop solution
  • Adhesive plate covers
Additional materials required
Protocol
Protocol Run time: 4 hours—30 minutes hands-on time.
Note: A standard curve must be run with each assay for quantification.
  1. Carefully read the product information sheet provided with the kit and create a plate layout (include blanks, standard, controls and sample).
  1. Allow all reagents to reach room temperature before use. Gently mix all liquid reagents prior to use.
  1. Add 100 µL of prepared standard and sample to the wells.
    Note: Samples prepared in Cell extraction buffer must be diluted 1:5 or greater in Diluent buffer. The dilution chosen should be optimized for each experimental system.
  1. Add 100 µL of diluted Detection antibody to the wells. Tap gently on the side of the plate to mix. Cover the plate with Adhesive plate cover and incubate at room temperature for 2 hours.
  1. Thoroughly aspirate the solution from the wells and discard the liquid.
  1. Wash the wells 4 times with 1X Wash buffer using a squirt wash bottle or an automated 96-well plate washer.
    Note: No matter your method of washing (squirt bottle or automated plate washer), fill wells with at least 400 µL of diluted wash buffer. Let it soak for 15–30 seconds, then decant the liquid. Tap the plate on clean absorbent paper to remove excess solution. Repeat. Use this method for all subsequent wash steps.
  1. Add 100 µL of diluted HRP conjugate to each well. Cover the plate and incubate at room temperature for 30 minutes.
  1. Thoroughly aspirate or decant the solution from the wells and discard the liquid.
  1. Wash the wells 4 times with 1X Wash buffer.
  1. Add 100 µL of Chromogenic substrate to each well. The substrate solution begins to turn blue.
  1. Incubate plate for 30 minutes at room temperature in the dark.
  1. Add 100 µL of Stop solution to each well. The solution in the wells changes from blue to yellow.
  1. Read the absorbance of each well at 450 nm. The plate must be evaluated within 30 minutes after adding the Stop solution.
  1. Use curve-fitting statistical software to plot a 4-parameter logistic curve fit to the standards and then calculate results for the test samples. See ELISA Data Analysis.
Note: If using an Antibody Pair Kit to build your own ELISA, you will need to coat the capture antibody onto a 96-well microplate yourself. For this process, you will require a 96-well microplate, capture antibody, and blocking buffer (usually BSA or milk diluted in PBS). Please follow these steps before starting the protocol above:
  1. Allow all reagents to reach room temperature before use. Gently mix all liquid reagents prior to use.
  1. Add 100 µL of diluted Capture antibody to each well. Cover plate and incubate at 4°C overnight.
  1. Bring the plate to room temperature. Thoroughly aspirate or decant solution from wells and discard the liquid.
  1. Wash the wells 4 times with Wash buffer.
  1. Add 200 µL of Blocking buffer to each well. Cover plate and incubate at room temperature for 1 hour.
  1. Thoroughly aspirate the solution from the wells and discard the liquid.
    --Continue with above protocol --
Microplate readers
Compare now
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.