Technical Resources - Media Formulations
N-2 Supplement (100X) liquid
For the growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype.
N-2 supplement is a chemically defined, 100X concentrate of Bottenstein's N-2 formulation (1). This supplement is recommended for the growth and expression of neuroblastomas and for the survival and expression of post-mitotic neurons in primary cultures from both the peripheral nervous system (PNS) and the central nervous system (CNS). It can be used as a substitute for the N-1 Bottenstein formulation. N-2 medium appears to be selective for neuronal cell lines and does not support the growth of nonneuronal cell lines. (in Current Methods in Cellular Neurobiology: Volume 4, edited by Jeffery Barker, John Wiley & Sons, Inc.,1983.)
ComponentsMolecular WeightConcentration (mg/L)mM
Proteins
Human Transferrin (Holo)78500.010000.00.12738854
Insulin Recombinant Full Chain5807.7500.00.0860926
Other Components
Progesterone314.470.630.0020033708
Putrescine161.01611.010.006211
Selenite173.00.520.0030057803
Verweis
1. Bottenstein, J.E. (1985) Cell Culture in the Neurosciences, Bottenstein, J.E. and Harvey, A.L., editors, p. 3, Plenum Press: New York and London.
Related Products: