Search
Contains no phenol red.
Neurobasal™ Media are basal media formulated to meet the neuronal cells special requirements. They allow for long-term maintenance of the normal phenotype and growth of neuronal cells, and maintain pure populations of neuronal cells without the need of an astrocyte feeder layer.
Catalog Number(s)
12348017| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 30.0 | 0.4 |
| L-Alanine | 89.0 | 2.0 | 0.02247191 |
| L-Arginine hydrochloride | 211.0 | 84.0 | 0.39810428 |
| L-Asparagine-H2O | 150.0 | 0.83 | 0.0055333334 |
| L-Cysteine | 121.0 | 31.5 | 0.2603306 |
| L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
| L-Isoleucine | 131.0 | 105.0 | 0.8015267 |
| L-Leucine | 131.0 | 105.0 | 0.8015267 |
| L-Lysine hydrochloride | 183.0 | 146.0 | 0.7978142 |
| L-Methionine | 149.0 | 30.0 | 0.20134228 |
| L-Phenylalanine | 165.0 | 66.0 | 0.4 |
| L-Proline | 115.0 | 7.76 | 0.06747826 |
| L-Serine | 105.0 | 42.0 | 0.4 |
| L-Threonine | 119.0 | 95.0 | 0.79831934 |
| L-Tryptophan | 204.0 | 16.0 | 0.078431375 |
| L-Tyrosine | 181.0 | 72.0 | 0.39779004 |
| L-Valine | 117.0 | 94.0 | 0.8034188 |
| Vitamins | |||
| Choline chloride | 140.0 | 4.0 | 0.028571429 |
| D-Calcium pantothenate | 477.0 | 4.0 | 0.008385744 |
| Folic Acid | 441.0 | 4.0 | 0.009070295 |
| Niacinamide | 122.0 | 4.0 | 0.032786883 |
| Pyridoxal hydrochloride | 204.0 | 4.0 | 0.019607844 |
| Riboflavin | 376.0 | 0.4 | 0.0010638298 |
| Thiamine hydrochloride | 337.0 | 4.0 | 0.011869436 |
| Vitamin B12 | 1355.0 | 0.0068 | 5.0184503E-6 |
| i-Inositol | 180.0 | 7.2 | 0.04 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
| Ferric Nitrate (Fe(NO3)3"9H2O) | 404.0 | 0.1 | 2.4752476E-4 |
| Magnesium Chloride (anhydrous) | 95.0 | 77.3 | 0.8136842 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 3000.0 | 51.724136 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 125.0 | 0.9057971 |
| Zinc sulfate (ZnSO4-7H2O) | 288.0 | 0.194 | 6.736111E-4 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 4500.0 | 25.0 |
| HEPES | 238.0 | 2600.0 | 10.92437 |
| Sodium Pyruvate | 110.0 | 25.0 | 0.22727273 |
NEUROBASAL™ Medium (1X) liquid
Cat No:
21103049
Neurobasal™-A Medium (1X) Liquid without Phenol Red
Cat No:
12349015
Neurobasal™-A Medium (1X) liquid
Cat No:
10888022
Neurobasal™-A Medium (1X) liquid, no D-glucose, no sodium pyruvate
Cat No:
A2477501