Looking for more physiologically relevant neuronal cultures? Discover Gibco™ Neurobasal™ PhysioPlus Medium, designed to support neuronal maintenance and functional activity.
Neurobasal™ Medium, minus phenol red
Gibco™
Neurobasal™ Medium, minus phenol red
Neurobasal Medium, minus phenol red is a basal medium designed for long-term maintenance and maturation of pure pre-natal and embryonicRead more
Have Questions?
Catalog NumberQuantity
12348017500 mL
Catalog number 12348017
Price (USD)
124.00
Each
Quantity:
500 mL
Customize this product
Price (USD)
124.00
Each
Neurobasal Medium, minus phenol red is a basal medium designed for long-term maintenance and maturation of pure pre-natal and embryonic neurons without the need for an astrocyte feeder layer when used with Gibco B-27 supplements. Use this modified formulation of Neurobasal Medium when the presence of phenol red will interfere with your results.
Neurobasal Medium is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of Neurobasal media and B-27 supplements.
For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Product LineNeurobasal
Product TypeNeurobasal Medium
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
Tested ForEndotoxin, Performance, Sterility, Osmolality, pH
ClassificationSerum-free
Culture TypeNeuronal Cell Culture
FormLiquid
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Frequently asked questions (FAQs)
Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?
No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What supplements should I use with Neurobasal or Neurobasal-A medium?
Neurobasal and Neurobasal-A media (for postnatal and adult neurons) allow for long-term maintenance of neuronal cells without the need for an astrocyte feeder layer. These media should be supplemented with either serum or a serum-free supplement, plus 0.5mM L-glutamine. B-27 supplement is a serum-free supplement that comes as a 50X concentrate in a 10ml volume. This is enough supplement for 500ml of media. Fetal, postnatal, and adult neural cultures can be grown in the appropriate Neurobasal medium supplemented with B-27 supplement .
We also have two other supplements. One is called G-5 and is for growth and expression of glial cells (normal and tumor) of astrocytic phenotype. This comes in a 1ml size, at a 100X concentration. The other supplement is called N-2 and is for growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype. This comes in a 5ml size, at a 100X concentration.
For more information on Neurobasal media, search "Neurobasal" from our website home page.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does the removal of phenol red from the Neurobasal Medium affect the culturing of neurons?
No, removal of phenol red from the Neurobasal Medium does not affect the culturing of neurons. Removal of phenol red allows culturing of neurons for studies involving certain receptors and downstream purification.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When should I use the Neurobasal Medium, minus phenol red?
We recommend using Neurobasal Medium, minus phenol red for studies involving certain receptors, downstream protein purification or other processes where the presence of phenol red is undesirable.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I understand that some media are worse than others for fluorescence imaging. How do I choose?
Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.