Search
Catalog Number(s)
11595030| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 650.0 | 8.666667 |
| L-Alanine | 89.0 | 225.0 | 2.52809 |
| L-Arginine hydrochloride | 211.0 | 700.0 | 3.3175356 |
| L-Asparagine | 132.0 | 350.0 | 2.6515152 |
| L-Aspartic acid | 133.0 | 350.0 | 2.631579 |
| L-Cystine | 240.0 | 22.0 | 0.09166667 |
| L-Glutamic Acid | 147.0 | 600.0 | 4.0816326 |
| L-Glutamine | 146.0 | 600.0 | 4.109589 |
| L-Histidine | 155.0 | 2500.0 | 16.129032 |
| L-Isoleucine | 131.0 | 50.0 | 0.3816794 |
| L-Leucine | 131.0 | 75.0 | 0.57251906 |
| L-Lysine hydrochloride | 183.0 | 625.0 | 3.4153006 |
| L-Phenylalanine | 165.0 | 150.0 | 0.90909094 |
| L-Proline | 115.0 | 350.0 | 3.0434783 |
| L-Serine | 105.0 | 550.0 | 5.2380953 |
| L-Threonine | 119.0 | 175.0 | 1.4705882 |
| L-Tryptophan | 204.0 | 100.0 | 0.49019608 |
| L-Tyrosine | 181.0 | 50.0 | 0.2762431 |
| L-Valine | 117.0 | 100.0 | 0.85470086 |
| beta-Alanine | 89.0 | 200.0 | 2.247191 |
| Vitamins | |||
| Biotin | 244.0 | 0.01 | 4.0983607E-5 |
| Choline chloride | 140.0 | 0.2 | 0.0014285714 |
| D-Calcium pantothenate | 477.0 | 0.02 | 4.192872E-5 |
| Folic Acid | 441.0 | 0.02 | 4.5351473E-5 |
| Nicotinic acid (Niacin) | 123.0 | 0.02 | 1.6260162E-4 |
| Para-Aminobenzoic Acid | 137.0 | 0.02 | 1.459854E-4 |
| Pyridoxine hydrochloride | 206.0 | 0.02 | 9.708737E-5 |
| Riboflavin | 376.0 | 0.02 | 5.319149E-5 |
| Thiamine hydrochloride | 337.0 | 0.02 | 5.934718E-5 |
| i-Inositol | 180.0 | 0.02 | 1.11111105E-4 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 750.0 | 6.756757 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 2280.0 | 11.231527 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 2780.0 | 11.300813 |
| Potassium Chloride (KCl) | 75.0 | 4100.0 | 54.666668 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 350.0 | 4.1666665 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 1013.0 | 7.3405795 |
| Other Components | |||
| Alpha-Ketoglutaric acid | 146.0 | 370.0 | 2.5342467 |
| D-Fructose | 180.0 | 400.0 | 2.2222223 |
| D-Glucose (Dextrose) | 180.0 | 700.0 | 3.8888888 |
| Fumaric acid | 116.0 | 55.0 | 0.47413793 |
| Malic acid | 134.0 | 670.0 | 5.0 |
| Succinic acid | 118.0 | 60.0 | 0.5084746 |
| Sucrose | 342.0 | 26680.0 | 78.011696 |