Search
Mouse intestine cryosection showing basement membranes labeled with chicken IgY anti-fibronectin antibody and Alexa Fluor® 488 Goat Anti–Chicken IgG antibody (A11039, green). Goblet cells and crypt cells were labeled with Alexa Fluor® 594 wheat germ agglutinin (W11262, red). The microvillar brush border and smooth muscle layers were visualized with Alexa Fluor® 680 phalloidin (A22286, pseudocolored purple). The section was counterstained with DAPI (D1306, D3571, D21490, blue).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›