Search
Bovine pulmonary artery endothelial cells (BPAEC) incubated with the fixable, mitochondrion-selective MitoTracker® Red CMXRos (M7512). After staining, the cells were formaldehyde-fixed, acetone-permeabilized, treated with DNase-free RNase and counterstained using SYTOX® Green nucleic acid stain (S7020). Microtubules were labeled with a mouse monoclonal anti–ß-tubulin antibody, biotin-XX goat anti–mouse IgG antibody (B2763) and Cascade Blue® NeutrAvidin biotin-binding protein (A2663). This photograph was taken using multiple exposures through bandpass optical filters appropriate for Texas Red® dye, fluorescein and DAPI using a Nikon Labophot 2 microscope equipped with a Quadfluor epi-illumination system.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Endothelial cells. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Nuclear, cytoskeletal, and mitochondrial staining of Muntjac fibroblasts. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Human tonsil section visualized with fluorescent Qdot® nanocrystal conjugates. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›