Search
A human tonsil section visualized with fluorescent Qdot® nanocrystal conjugates. Ki67 in the nuclei of proliferating B-cells was labeled with a rabbit anti-Ki67 primary antibody and visualized using red-fluorescent Qdot® 585 goat F(ab')2 anti-rabbit IgG (Q11411MP). Keratin in the peripheral stromal cells was labeled with a mouse anti-pan-keratin monoclonal antibody and visualized using a biotin-goat anti-mouse IgG followed by a green-fluorescent Qdot® 525 streptavidin conjugate (Q10141MP). Nuclei were stained with blue-fluorescent Hoechst 33342 (H1399, H3570, H21492).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Endothelial cells. Go ›
Three-color staining of HeLa cells using fluorescent Qdot® nanocrystal conjugates. Go ›
Porcine primary skeletal muscle cell transduced with Organelle Lights fluorescent proteins. Go ›