Multicolor fluorescence image of mouse intestine cross section.
Multicolor fluorescence image of mouse intestine cross section. Filamentous actin prevalent in the brush border was visualized with Alexa Fluor® 568 phalloidin (Cat. no. A12380). Goblet cell mucus was detected with a blue-fluorescent Alexa Fluor® 350 conjugate of wheat germ agglutinin (Cat. no. W11263). Nuclei were stained with SYTOX® Green nucleic acid stain (Cat. no. S7020).
Multicolor fluorescence image of mouse intestine cross section. Filamentous actin prevalent in the brush border was visualized with Alexa Fluor® 568 phalloidin (Cat. no. A12380). Goblet cell mucus was detected with a blue-fluorescent Alexa Fluor® 350 conjugate of wheat germ agglutinin (Cat. no. W11263). Nuclei were stained with SYTOX® Green nucleic acid stain (Cat. no. S7020).
Related Products
Related Images