Search
HeLa cell transfected with pShooter pCMV/myc/mito/GFP, then fixed and permeabilized. Green-fluorescent protein (GFP) localized in the mitochondria was labeled with anti-GFP mouse IgG2a (Cat. no. A11120) and detected with orange-fluorescent Alexa Fluor® 555 goat anti–mouse IgG (CAt. no. A21422), which colocalized with the dim GFP fluorescence. F-actin was labeled with green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379), and the nucleus was stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490). The sample was mounted using ProLong® Gold antifade reagent (Cat. no. P36930). Some GFP fluorescence is retained in the mitochondria after fixation (top), but immunolabeling and detection greatly improve visualization (bottom).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Live cell imaging with CellLight™ reagents. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›