Cultured axolotl sensory neurons stained with our anti–human neuronal protein HuC/HuD antibody and detected using the red-orange–fluorescent Alexa Fluor® 546 goat anti–mouse IgG antibody.
Cultured axolotl sensory neurons stained with our anti–human neuronal protein HuC/HuD antibody (anti-Hu, Cat. no. A21271) and detected using the red-orange–fluorescent Alexa Fluor® 546 goat anti–mouse IgG antibody (Cat. no. A11003). Actin was labeled using green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379) and nuclei were stained with blue-fluorescent Hoechst 33342 (Cat. no. H1399, H3570, H21492). The image was contributed by Josh Gross and Linda Barlow, University of Denver, Denver, Colorado.
Cultured axolotl sensory neurons stained with our anti–human neuronal protein HuC/HuD antibody (anti-Hu, Cat. no. A21271) and detected using the red-orange–fluorescent Alexa Fluor® 546 goat anti–mouse IgG antibody (Cat. no. A11003). Actin was labeled using green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379) and nuclei were stained with blue-fluorescent Hoechst 33342 (Cat. no. H1399, H3570, H21492). The image was contributed by Josh Gross and Linda Barlow, University of Denver, Denver, Colorado.
Related Products
Related Images