Live-cell labeling using CellMask™ Deep Red plasma membrane stain.
Live-cell labeling using CellMask™ Deep Red plasma membrane stain. HaCaT cells (an immortalized human keratinocyte cell line) transfected with a mitochondrion-targeted eGFP construct were incubated for 15 minutes in a solution containing 1 µM Hoechst 33342 (Cat. no. H21492) and then for 5 minutes in a fresh solution containing 8 µg/ml CellMask™ Deep Red plasma membrane stain (Cat. no. C10046); staining took place at 37°C in the presence of 5% CO2. After washing twice with DMEM (including 10% FBS) at 37°C, the cells were imaged with a Zeiss Cell Observer HS microscope fitted with a 100× oil alpha Plan-Apochromat objective and a BFP/GFP/HcRed filter set from Zeiss (62HE). The image stack was deconvolved with the Zeiss AxioVision 3D Deconvolution module with the point spread function acquired using a 200 nm TetraSpeck™ fluorescent microsphere (Cat. no. T7280). Metaphase chromosomes (pseudocolored blue, Hoechst 33342), plasma membrane (pseudocolored red, CellMask™ Deep Red plasma membrane stain), and mitochondria (pseudocolored green, eGFP) are all clearly visible. Image submitted by Christian Junker, University of Saarland, Department of Biophysics, Homburg, Germany.
Live-cell labeling using CellMask™ Deep Red plasma membrane stain. HaCaT cells (an immortalized human keratinocyte cell line) transfected with a mitochondrion-targeted eGFP construct were incubated for 15 minutes in a solution containing 1 µM Hoechst 33342 (Cat. no. H21492) and then for 5 minutes in a fresh solution containing 8 µg/ml CellMask™ Deep Red plasma membrane stain (Cat. no. C10046); staining took place at 37°C in the presence of 5% CO2. After washing twice with DMEM (including 10% FBS) at 37°C, the cells were imaged with a Zeiss Cell Observer HS microscope fitted with a 100× oil alpha Plan-Apochromat objective and a BFP/GFP/HcRed filter set from Zeiss (62HE). The image stack was deconvolved with the Zeiss AxioVision 3D Deconvolution module with the point spread function acquired using a 200 nm TetraSpeck™ fluorescent microsphere (Cat. no. T7280). Metaphase chromosomes (pseudocolored blue, Hoechst 33342), plasma membrane (pseudocolored red, CellMask™ Deep Red plasma membrane stain), and mitochondria (pseudocolored green, eGFP) are all clearly visible. Image submitted by Christian Junker, University of Saarland, Department of Biophysics, Homburg, Germany.
Related Products
Related Images