Search
Potential-dependent staining of mitochondria in CCL64 fibroblasts by JC-1 (T3168). The mitochondria were visualized by epifluorescence microscopy using a 520 nm longpass optical filter. Regions of high mitochondrial polarization are indicated by red fluorescence due to J-aggregate formation by the concentrated dye. Depolarized regions are indicated by the green fluorescence of the JC-1 monomers. The image was contributed by Lan Bo Chen, Dana Farber Cancer Institute, Harvard Medical School.
Agarose gel containing camptothecin-treated HL-60 cells. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›