Search
Mitotic divisions in early Drosophila embryos. The spindles were labeled with an anti–a-tubulin primary antibody and probed with a secondary antibody labeled with Lissamine rhodamine B sulfonyl chloride (Cat. No. L20, L1908). After a brief RNase treatment to reduce background fluorescence, the chromosomes were counterstained with SYTOX® Green nucleic acid stain (Cat. No. S7020). The image was contributed by Tulle Hazelrigg and Amy MacQueen, Columbia University.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Endothelial cells. Go ›