Search
Fixed and permeabilized osteosarcoma cells simultaneously stained with the fluorescent lectins Alexa Fluor® 488 concanavalin A (Con A) (Cat. no. C11252) and Alexa Fluor® 594 wheat germ agglutinin (WGA) (Cat. no. W11262). Con A selectively binds a-glucopyranosyl residues, whereas WGA selectively binds sialic acid and N-acetylglucosaminyl residues. The nuclei were counterstained with blue-fluorescent Hoechst 33342 nucleic acid stain (Cat. no. H1399, H3570, H21492). The image was acquired using bandpass filter sets appropriate for the Texas Red® dye, fluorescein and DAPI.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›