Search
Endogenous alkaline phosphatase activity of osteoblast cells in a cartilaginous element of an adult zebrafish head cryosection. The activity was localized with the ELF 97 Endogenous Phosphatase Detection Kit (E6601). In addition to the yellow-green fluorescence of the ELF 97 precipitate, the section was stained with Texas Red-X wheat germ agglutinin (W21405) and counterstained with the Hoechst 33342 nucleic acid stain (H1399, H3570, H21492). The triple-exposure image was acquired using bandpass filter sets appropriate for the ELF precipitate, Texas Red dye and AMCA.
Prostate carcinoma. Go ›
Transformed mouse fibroblasts (CRE BAG 2 cells). Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›