Search
FluoCells® prepared slide #4 (Cat. no. F24631) contains a section of mouse intestine stained with a combination of fluorescent stains. Alexa Fluor® 350 wheat germ agglutinin (Cat. no. W11263) is a blue-fluorescent lectin that was used to stain the mucus of goblet cells. The filamentous actin prevalent in the brush border was stained with red-orange–fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Finally, the nuclei were stained with SYTOX® Green nucleic acid stain (Cat. no. S7020). This image is a composite of three digitized images obtained with filter sets appropriate for fluorescein, DAPI and tetramethylrhodamine.
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Developing Drosophila embryo Go ›
Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate Go ›