Search
Capillaries in the hippocampal region of a mouse brain cryosection were visualized with the green-fluorescent Alexa Fluor® 488 conjugate of lectin HPA from Helix pomatia (Cat. No. L11271), which specifically binds to type A erythrocytes and a-N-acetylgalactosaminyl residues. The nuclei were counterstained with nuclear yellow (Cat. No. N21485). The multiple-exposure image was acquired using a DAPI longpass filter set and a filter set appropriate for fluorescein.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Endothelial cells. Go ›