Search
A 2.0 µm maize leaf section illustrating the immunolocalization of the enzyme ribulose bisphosphate carboxylase (rubisco) in the chloroplasts of the bundle sheath cells surrounding the vascular bundles. Maize is a C4 plant and, as a result, spatially segregates components of the photosynthetic process between the leaf mesophyll and the bundle sheath. Rubisco was localized using a rabbit anti-rubisco antibody and visualized using the highly cross-adsorbed Alexa Fluor® 488 goat anti–rabbit IgG antibody (Cat. No. A11034). The remaining fluorescence is due to the autofluorescence of chlorophyll, which appears red and is localized to the mesophyll plastids; lignin, which appears dull green and is localized to the xylem of the vascular bundle; and cutin, which appears bright green and is localized to the cuticle outside the epidermis. Image contributed by Todd Jones, DuPont.
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›