Search
Bovine pulmonary artery endothelial cells (BPAEC) incubated simultaneously with 50 nM LysoTracker® Red DND-99 (Cat. no. L7528) and 75 nM MitoTracker® Green FM (M7514) at 37°C for 30 minutes. Both dyes showed excellent cellular retention, even after cells were fixed in 3% glutaraldehyde for 30 minutes. The image was deconvolved using Huygens software (Scientific Volume Imaging, http://www.svi.nl/).
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›