Search
Fixed and permeabilized muntjac skin fibroblasts stained with Alexa Fluor 488 phalloidin (A12379), an anti–a-tubulin antibody (A11126) and anti–cdc6 peptide antibody. The anti–a-tubulin antibody was prelabeled with the Zenon Alexa Fluor 568 Mouse IgG1 Labeling Kit (Z25006), and the anti–cdc6 peptide antibody was prelabeled with the Zenon Alexa Fluor 350 Mouse IgG1 Labeling Kit (Z25000).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Mouse anti-ATP Synthase Subunit IF1 Monoclonal Antibody. Go ›