Search
Welcome to our Solutions section, where we offer advanced technologies for flow measurement that are tailored to meet the diverse needs of the industrial and manufacturing sectors. At Thermo Fisher Scientific, we understand the critical importance of precise and reliable flow measurement in optimizing your processes and ensuring operational efficiency. Our comprehensive range of flow computers solutions utilizes advanced technology that delivers exceptional accuracy, providing you with the tools necessary to enhance productivity, help ensure safety, and achieve your performance goals. Explore our offerings to find the perfect solution for your specific application needs.
Thermo Scientific AutoConfig Software is designed to simplify the setup and configuration of flow measurement systems. This intuitive software streamlines the process of calibrating and maintaining flow meters, reducing the time and expertise required for system setup. It offers user-friendly interfaces and automated workflows to enhance accuracy and efficiency in flow measurement, ensuring reliable and precise data for industrial applications.
Designed to empower the user, the interface is simple to configure eliminating the need for in depth programming.
AutoCONFIG Software is the robust interface for the Thermo Scientific AutoSERIES of Flow Computers. Designed to empower the user, the interface is simple to configure and eliminates the need for in-depth programming.
The flow computer series provides advanced solutions for accurate hydrocarbon measurement in the oil and gas industry. These flow computers are designed to handle a wide range of applications, from custody transfer to process control, ensuring precise and reliable data. The AutoSeries offers user-friendly interfaces, robust data handling, and seamless integration with existing systems, making them ideal for enhancing operational efficiency and regulatory compliance in hydrocarbon measurement.
From field to control room and upstream to downstream, our products provide greater control, confidence, and reliability.
Transport, control, and store your product reliably with the aid of the Thermo Scientific flow computer series. From a single measurement point to large delivery points, the flow computer, coupled with the AutoCONFIG interface, helps oil and gas transporters maximize flow throughput with quick field deployment and enhanced station control.
This selection guide is designed to help you choose the ideal flow computers solutions for your oil and gas operations. Simplify your decision-making process and optimize your operations with this essential resource.
Use this comparison chart to see which flow computer is right for you.
Thermo Scientific flow computers incorporate the latest measurement standards and calculations for hydrocarbon measurement. Use this comparison chart to see which flow computer is right for you.
| AutoFLEX Scalable All-in-One Flow Computer Platform | AutoPILOT PRO Multi-Run Hydrocarbon Flow Computer | ||
![]() |
![]() |
![]() |
|
Run Capacity |
up to 32 | 1-2 Runs |
1-6 Runs |
Plunger Capability |
Yes | Yes | Yes |
API 21.1 Compliant |
Yes | Yes | Yes |
Field Mountable |
Yes** | Yes | Yes |
API 21.1 & 21.2 Compliant |
Yes | Yes | Yes |
Expandable / Multiselect IO |
Yes | Expandable IO Only |
Expandable IO Only |
GC Connectivity |
Yes | Yes |
Yes |
Full Station Control |
Yes | No |
Yes |
Rack Mount |
Yes | No |
No |
USM Diagnostics |
Yes | No |
Yes |
| Bluetooth | Yes | Yes |
No |
Intrinsically Safe |
No | No |
Yes |
Proving Loop Control |
Yes | No |
No |
Custom Reporting |
Yes | Yes |
Yes |
Flow Calculation Speed |
1x Sec | 1x Sec |
1x Sec |
**AutoFLEX 3 with Enclosure only