ALPS5000 Automated Plate Heat Sealer
ALPS5000 Automated Plate Heat Sealer
ALPS5000 Automated Plate Heat Sealer
Thermo Scientific™
ALPS5000 Automated Plate Heat Sealer
Fast, reliable and quiet automated plate sealing solution for any lab.
Have Questions?
Catalog NumberVoltage
AB-5000110/240 V
Catalog number AB-5000
Price (USD)
39,784.50
Online Exclusive
46,415.00
Save 6,630.50 (14%)
Each
Voltage:
110/240 V
Price (USD)
39,784.50
Online Exclusive
46,415.00
Save 6,630.50 (14%)
Each
The Thermo Scientific ALPS5000 is the next generation in plate sealing equipment offering fast sealing times, increased process customization and all electric operation improving reliability and reducing noise.
  • Electrically Driven with Tool Free Loading
    • No need to connect to an external air source or replace expensive vacuum cups
    • Reduced risk of contamination inherent in pneumatically driven sealers
    • Quieter operation when compared to pneumatically driven systems
    • Tool free loading makes reloading foil simple and repeatable
  • Touchscreen Control and Savable Protocols
    • Scalable to meet labs needs now and into the future
    • Precise control over sealing parameters to ensure repeatability
    • Easily integrated into automation or robotics workflows
  • Sealing of Multiple Plate Heights
    • Seal plates from 8mm to 46mm in height with no adapters required
    • Compatible with high throughput automated applications
Specifications
Certifications/ComplianceUL/NRTL, CE
DescriptionAutomated Plate Heat Sealer
Dimensions (L x W x H)19 x 9 x 21 in.
Electrical Requirements110/240 V, 50/60 Hz
Height (English)21 in.
Height (Metric)525 mm
Hertz50/60 Hz
Length (English)19 in.
Length (Metric)482.6 mm
No. per Pack1 Ea.
Sealing Temperature RangeAmbient to 199°C
Voltage110/240 V
Weight (English)55 lb.
Weight (Metric)25 kg
Width (English) 9 in.
Width (Metric)228 mm
Product TypePlate Sealer
Unit SizeEach