AutoXP Gas and Liquid Flow Computer
AutoXP Gas and Liquid Flow Computer
Thermo Scientific™
AutoXP Gas and Liquid Flow Computer
The AutoXP Gas and Liquid Flow Computer is a flexible unit that can be used as a single-run flow computer or smart multi-variable transmitter for both gas and liquid applications.
Have Questions?
Catalog number AUTOXP
Price (USD)
-
Request A Quote
Gain ultimate flexibility with the Thermo Scientific™ AutoXP Gas and Liquid Flow Computer. The AutoXP unit can be used as a single-run flow computer or smart multi-variable transmitter. This system also can be configured for both gas and liquid applications utilizing today's most common primary devices all while meeting Class I Div 1 requirements and API 21.1 (custody transfer standard).
The AutoXP instrument is built on the field-proven AutoCONFIG platform. Designed for harsher environments across all oil and gas sectors, the flow computer is configurable MODBUS™ Protocol mapping for a customizable user interface.
  • Rugged, robust unit for well testing
  • Ease of connectivity out in the field via Bluetooth™
  • Preprogrammed functions simplify automation
  • Through glass keypad interface for enhanced worker safety
  • Designed to meet Class I Div 1 requirements
AutoCONFIG Configuration Software
Thermo Scientifiic flow computers are built on an innovative field proven platform incorporating the latest measurement standards and calculations for hydrocarbon measurement. AutoCONFIG interface allows for simplified configuration eliminating need for indepth programming. Control functions include Station Control, PID, Alarming, Event-based Logging and many more.
Specifications
Additional Factors/EquationsFWV (manual, partial or full); FWS:NIST14
Audit TrailsUser configurable, defaulting to 200 audit events
Certifications/ComplianceCSA/C-US Class I, Div 1, Groups B, C, D; ambient temperature range of -40°C to +85°C (-40°F to +185°F), temperature code T6 (-40°C to 75°C) T5 (-40°C to 85°C) EN 61326-1: 2013 (Industrial Criteria); FCC 47 CFR Part 15, Subpart B; ICES 003: 2016; Measurement Canada, Approval no. AG-0650
DiagnosticAGA 10 SoS
Differential Meters(DP, Orifice) AGA 3/ANSI/API 2530-1992 Method 2; AGA 3/ANSI/API 2530-1985; ISO 5167; Cone meters; Annubar; GOST
Display128x65 backlit LCD display; User programmable scroll list and menus
EnergyAGA 5; GPA 2172; ISO 6976
Ethylene Density CalculationAPI 2565 (Ch 11.3.2.1); Ethylene NBS 1045
Keypad4 IR sensors through glass key input
Linear Meters(Turbine) AGA 7; AGA 9; AGA 11
No. of AlarmsUser configurable, defaulting to 200 alarm events
Operating Humidity Range0 to 95% RH, Non-condensing
Operating Temperature Range-40°C to +85°C (-40°F to +185°F)
Processor32 bit
Product LineAutoXP
Propylene Density CalculationAPI Ch 11.3.3.2
Supercompressibility(Fpv) AGA 8 Gross-1992; AGA 8 Detail-1992; AGA 8 Short-1988; NX-19; NX-19 Analysis; GERG
Turbine Meter Linearization10 Point Frequency/K-factor Table
Volume Correction FactorConsistent with API 2540/ASTM D1250-80/IP 200; 5/6 A/B; 23/24 A/B; 53/54 A/B; 6/24/54 C; CH 11.1 2004; Note: natural gas liquids (NGL) and liquefied petroleum gases (LPG): OLD 23/24, OLD 53/54; Table E is new standard to replace OLD 23/24
Data Memory2 MB SRAM
Enclosure RatingIP67 (NEMA 4X)
Input Power10 VDC to 30 VDC
Live Density InputUGC, 4 to 20 mA
Product TypeGas and Liquid Flow Computer
Program Memory4 MB Flash
Unit SizeEach