CEDIA™ Buprenorphine II Drugs of Abuse Assay
Actual product may vary
Thermo Scientific™
CEDIA™ Buprenorphine II Drugs of Abuse Assay
Determine the presence of Buprenorphine and all major metabolites with the new, improved formulation of this immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100208501 x 65 mL, MCC
100208493 x 17 mL, TSC
Catalog number 10020850
Price (USD)
-
Quantity:
1 x 65 mL, MCC
Determine the presence of Buprenorphine and its metabolites with the new, improved Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Assay. It is the only assay currently available on the market which detects all three major metabolites: norbuprenorphine, buprenorphine-3-β-glucuronide and norbuprenorphine-3-β-glucuronide. Buprenorphine is widely used as a practical alternative to methadone in the treatment of opioid addiction. This assay has no significant cross-reactivity to other opioids and does not cross-react with morphine, even at 100,000ng/mL.
  • Detect Buprenorphine and its metabolites: buprenorphine-3-β-glucuronide, norbuprenorphine, and norbuprenorphine-3-β-glucuronide
  • Qualitative or semi-quantitative options for results
  • No cross-reactivity with any other opioids
  • Applications available on a wide range of clinical chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsCEDIA Buprenorphine II Control Set
DescriptionCEDIA Buprenorphine II Assay
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable AnalytesBuprenorphine (BUP), Norbuprenorphine (NBUP), Buprenorphine-3-β-glucuronide, Norbuprenorphine-3-β-glucuronide
DoA CalibratorsBuprenorphine II Calibrator, Negative Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C