Specifications
AutoclavableNot Autoclavable
Closure SizeID x wall: 0.32 cm x 0.16 cm (1/8 in. X 1/16 in.)
Closure TypeWeldable (TPE) Tubing, Luer-Lock female 1/8 in.
DescriptionCentriPAK BPC Single
For Use With (Application)For mammalian, yeast and bacterial cell separations
For Use With (Equipment)Sorvall BIOS 16 Bioprocessing Centrifuge
Includes2D BPC Chamber
Max. Sample Density1.2 g/mL
Max. Temperature (English)104°F
Max. Temperature (Metric)40°C
PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Product LinePolaris
QuantitySet of 2
Shipping ConditionPackages placed in strong cardboard cartons with product and lot identification
TransparencyTranslucent
Capacity (English)57.48 oz.
Capacity (Metric)1700 mL
Certifications/ComplianceUSP<88>, USP<87>, USP<661>, EP<3.2.2.1>, USP<85>, USP<788>, EMEA/410/01
DisposableSingle-use
GraduatedNo
MaterialCX3-9 Film
Max. Force7187 x g
Product TypeFlexible Bioprocess Container
SterilitySterile
Unit SizeEach