Charge Variant Analysis
Meeting your needs for robust charge variant profiling
Biotherapeutic proteins such as monoclonal antibodies (mAbs) are far more heterogeneous than small-molecule drugs. The presence of the charged state can significantly impact the structure, stability, binding affinity, and efficacy of the biotherapeutic drug. It is therefore necessary to understand the profile of the drug so that charge variants are identified and removed if necessary.
The heterogeneity of monoclonal antibodies is revealed by charge sensitive methods; such as ion exchange chromatogram (IEX). We offer a range of buffers, ion exchange or hydrophobic interaction column chemistries, and UHPLC instrumentation to meet the needs for robust analysis of charge variants in monoclonal antibodies and therapeutic proteins.
Contact us  Receive updates
Charge variant profiling workflow
Charge variant profiling workflow
Charge variant analysis categories
Achieve robust and reproducible gradients using Thermo Scientific pH Gradient Buffer platform. They are applicable to a wide range of mAbs and ready to use with existing LC columns and systems—including the Thermo Scientific Vanquish Flex UHPLC system—without the need to formulate mobile phases. The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of mAb charge variants through a generic LC-based approach to mAb characterization.
Columns for separating protein charge variants
The gold standard column for charge variant profiling of monoclonal antibodies is widely regarded  as the Thermo Scientific ProPac WCX-10. ProPac columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions. The cation-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
We developed our next generation cation exchange column to extend column lifetime and method robustness - the Thermo Scientific MAbPac SCX-10 LC Column is for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants. A hydrophilic layer surrounds polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Proven Charge Variant Workflow at global biopharma CRO
Speed and accuracy are key criteria for laboratories performing charge variant profiling. Especially in contract research organizations who deliver confident, fast analytical services to the biopharmaceutical and biosimilar industry. Laboratories like Sartorius Stedim BioOutsource have created a niche within the industry, dedicated to rapid structural evaluation of biosimilar monoclonal antibodies (mAbs). Hear from BioOutsource about how the Thermo Scientific Charge Variant Workflow allows them to easily and robustly analyze a range of mAbs using a single platform ph gradient IEX method.
 Read case study
Chromatography system built for robust routine protein characterization
Obtain new benchmarks in accuracy, precision, and sensitivity in a biocompatible UHPLC system with state-of-the-art quaternary solvent blending. The Thermo Scientific Vanquish Flex Quaternary UHPLC system shares all the values inherent to the Vanquish product line, including an innovative design, focus on uptime, robustness, and reliability. Multiple detection options as well as Thermo Scientific Viper Fingertight Fitting connections throughout the system give you the performance you need with the ease of use you want. The combination makes this system ideal for biopharmaceutical and pharmaceutical applications.
Chromatography data solution for GMP testing of protein charge variants
Streamline your laboratory workflow using Thermo Scientific Chromeleon 7.3 CDS software. This software delivers superior instrument control, automation, and data processing for compliant GxP biopharmaceutical manufacturing and QA/QC environments.
Run your routine HPLC, UHPLC, or LC-MS protein charge variant analyses in an enterprise environment—from method creation to final reporting.
Featured videos
Application - The product is For General Lab Use Only - Not For Diagnostic Procedures
Our General Purpose product-lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks.
The product is For General Lab Use Only - Not For Diagnostic Procedures. The application is For Research Use Only. Not for use in diagnostic procedures.