TriPlus™ RSH EQuan 850 System for Water Testing
TriPlus™ RSH EQuan 850 System for Water Testing
TriPlus™ RSH EQuan 850 System for Water Testing
TriPlus™ RSH EQuan 850 System for Water Testing
Thermo Scientific™
TriPlus™ RSH EQuan 850 System for Water Testing
Screen for trace contaminants in water and beverages quickly, easily, and with greater confidence.
Have Questions?
Catalog number C0950-01-00614
Price (USD)
-
Exposure to pesticides, pharmaceuticals, personal care products, endocrine disruptors, and perfluorinated compounds in water is a significant health concern. The Thermo Scientific™ TriPlus™ RSH EQuan 850 system for water testing provides a turn-key solution for screening, identification, and quantitation of trace contaminants in environmental water, drinking water and beverage samples. It also automates the preparation of calibration standards and solid phase extractions.
Improve laboratory productivity
  • Online sample preparation reduces analytical variability
  • Column switching enables the automation of sample preconcentration
  • Automatic Tool Change allows large volume injections without user intervention, and the use of different syringes within the same analytical method
  • Ultra-high-pressure LC (UHPLC) pump enables fast separations of complex samples
Exceptional flexibility
The TriPlus RSH EQuan 850 system is compatible with several Thermo Scientific mass spectrometers:
  • Thermo Scientific™ TSQ™ series triple quadrupole mass spectrometers for the ultimate sensitivity, precision and accuracy in quantitative workflows
  • Thermo Scientific™ Q-Exactive™, Thermo Scientific™ Orbitrap Exploris™ and Thermo Scientific™ Orbitrap ID-X™ Tribrid™ series mass spectrometers combine full-scan MS/MS, superior resolution and mass accuracy to ensure the highest possible confidence in every ID.
Specifications
DescriptionLC TriPlus RSH EQuan 850 system for water analysis
Injection Volume RangeUp to 20 mL
Temperature Range4°C - 40°C
Injection TypeOnline SPE and standard Injection
Sample Capacity6x well-plates (96 and 384); 6x54 2 mL vials; 6x15 10 mL vials
Unit SizeEach