Orbitrap Exploris 120 Mass Spectrometer
The new standard in productivity
The proven Thermo Scientific Orbitrap technology in the Thermo Scientific Orbitrap Exploris 120 mass spectrometer accelerates your route to qualitative and quantitative confidence for both method development and everyday testing purposes. With its unique ability to provide consistently accurate data and a design made for operational simplicity, it sets the new standard in instrument productivity and ruggedness.
On this page
Introducing the Orbitrap Exploris 120 mass spectrometer
Hear directly from the Veterinary Diagnostic Laboratory at Iowa State University about how the Orbitrap Exploris 120 mass spectrometer enables them to provide high quality service to their clients who quickly need results in a variety of sample types.
Key benefits of the Orbitrap Exploris 120 mass spectrometer
Confidently perform high-throughput qualitative and quantitative analysisKeep your laboratory running seamlessly with the new standard in instrument ruggednessFocus on driving results with faster method set-up
Spend less time questioning your results with the unique ability of Orbitrap high-resolution, accurate-mass spectrometers (HRAM) to provide accurate results and reduce false positives or negatives. Confidently resolve ions of interest from interfering ions with high mass resolution up to 120,000 at m/z 200 for both method development and everyday testing analyses.Maximize return on your investment with capabilities that minimize downtime. Perform a single, stable calibration for the entire mass range from m/z 40-3000. With an improved scan rate up to 22 Hz, maximize your compound coverage with confidence. The ability to do both positive and negative mode switching ensures you analyze twice as many analytes in the same sample.Empower high-resolution MS users by giving them the ability to analyze samples using ready-to-run method templates. Minimize time spent on software training for Thermo Scientific MS users with a common software user interface for data acquisition.
How the Orbitrap Exploris 120 MS works
Discover how the Orbitrap Exploris 120 MS instrument enables you to prioritize your time on making critical decisions which impact the future of you or your client's organization. The latest instrument capabilities have been designed with the sole purpose of putting you on the fast path to accurate, reportable results. Leave the lab at the end of the day feeling productive and accomplished.
New high-field Orbitrap mass analyzer increases resolving powerBoost data quality with Advanced quadrupole technology (AQT)Ultimate ruggedness with Thermo Scientific OptaMax NG ion source
Increased resolution and speed up to 120,000 resolution and 22 Hz scan speed.MS/MS precursor ion selection with high transmission from m/z 40 to 3,000.Efficient, heated electrospray ionization (H-ESI II) probe to deliver maximum performance with minimal optimization. You do have the option of the APCI probe which can be upgraded to APPI.
Orbitrap Exploris 120 etozazole isopyrazam spectra
Full-scan mass spectrum of Etoxazole and Isopyrazam, measured using the Orbitrap Exploris 120 mass spectrometer at a resolution (R) setting up to 120,000 (FWHM) at m/z 200, demonstrates outstanding resolving power.
Access sub-ppm mass accuracy for at least five days with the Thermo Scientific EASY-IC ion source. With automated internal calibrant delivery, the EASY-IC ion source saves you time so you can quickly return to running your samples. Use the Thermo Scientific Pierce FlexMix calibration solution to perform a single calibration for the full mass range of m/z 40–3000, in both positive and negative modes.
Orbitrap Exploris 120 easy LC internal calibration
Through the use of Thermo Scientific EASY-IC ion source, sub-ppm mass accuracy is consistently delivered without user-intervention for at least five days. A panel of veterinary drugs with m/z range from 160…900 was analyzed at 100 µg/L over a 15-day period with continual polarity switching, providing exceptional mass accuracy.
The Orbitrap Exploris 120 mass spectrometer has demonstrated its ability to consistently deliver desired results on multiple instruments.
Orbitrap system consistency
To assess LC-HRAM MS system-to-system reproducibility, two operators at two different locations analyzed the same sample of veterinary drug residues in bovine muscle matrix on two separate systems. The minimal 5.1% average difference observed in analyte concentrations demonstrates high reproducibility across both Orbitrap Exploris 120 mass spectrometer systems.
Orbitrap Exploris 120 peak area
Long term system robustness demonstrated for pesticides in olive oil over 250 injections with polarity switching.
The Orbitrap Exploris 120 mass spectrometer triggers even more precursor ions for MS/MS with an improved scan rate of up to 22 Hz so you can turn around sample results confidently. If you want to run shorter UHPLC gradients, you can do so with excellent resolution, sensitivity and mass accuracy.
Orbitrap Exploris 120 MS fast scan rate
Quantitation of all compounds in bovine muscle matrix is easily achieved by fast scan speeds, even when multiple precursor ion m/z isolation windows are selected for the DIA experiment.
The Orbitrap Exploris 120 mass spectrometer enables positive and negative mode switching with a single cycle completed in approximately 1.4 Hz. Compared to other high-resolution MS instruments, the mass resolution and mass stability do not need to recover after polarity switching, saving you time.
Orbitrap Exploris 120 MS rapid polarity switching
Full scan MS data acquired for 90 ng/g Zearalenone in corn feed demonstrates preservation of mass accuracy stability during polarity switching.
Being able to focus on your science, rather than experimental set-up through ready-to-use application-specific method templates, means that the next breakthrough is closer than ever before. The Thermo Scientific Method Editor and Tune ecosystem deliver intuitive method programming with pre-defined (yet fully editable) templates for the most popular experiments.
MethEdit Orbitrap Exploris 480 Flow
Move from method set up to analysis quickly using ready-to-use application-specific templates.
If you’re an experienced triple quadrupole MS user, performing quantitation on an Orbitrap HRAM MS instrument will require the use of different scan modes. There are multiple scan modes, but listed below are the most popular ones. Keep in mind the importance of specifying narrow mass ranges especially if your analyses require increased sensitivity or selectivity. The quadrupole excludes any ions that are not in the user-defined mass range enabling higher quality results.
  • Full Scan MS – most predominantly used due to comprehensive, high-quality quantitative data provided. Once a mass range is defined to capture all relevant ions, rest assured detection of all compounds in that range will be measured.
  • Targeted Selected Ion Monitoring (t-SIM) – used for analyses requiring higher sensitivity because you can set a narrow mass range (as narrow as 0.4 Da).
  • Targeted -MS2 (t-MS2 formerly Parallel Reaction Monitoring (PRM)) – used for analyses in complex samples requiring additional selectivity. A fragmentation-based acquisition mode, Targeted-MS2 provides more selectivity because a high-resolution product ion spectrum is created for only desired precursor ions based on the inclusion list.
How to select the best Orbitrap mass spectrometer scan mode for your application
Free on-demand webinar: Multi-residue analysis in complex matrices: Selecting the best Orbitrap MS scan mode for your application
Data analysis software: Streamline small molecule qualitative and quantitative assays
Connecting small molecule unknown identification and characterization through to screening and quantitation doesn’t just stop at data acquisition across multiple platforms. Now that you have acquired comprehensive, high-quality MS and MS2 data, you need to identify and characterize unknowns and transfer discovery knowledge through to every day testing.
Which data analysis software is right for you?
NeedsCompound Discoverer softwareTraceFinder softwareChromeleon CDS software
Non-targeted screening (unknown knowns and unknown unknowns)
Targeted screening (known unknowns)
Non-targeted and targeted quantitation
Targeted quantitation only (known knowns)
Complex targeted panel analysis
GxP/21 CFR Part 11 compliance capable
Library compatibility
Thermo Scientific Chromatography and source compatibility
Third Party Instrument Control
= Not applicable = Poor = Good = Better = Best
Our connected data processing software ecosystem streamlines interpretation of your high-confidence discovery data through powerful Thermo Scientific Compound Discoverer software with direct access to the world’s largest LC-MSn reference spectral library, Thermo Scientific mzCloud, or through utilization of offline or proprietary libraries. These libraries act as your knowledge repository and allow direct transfer through to everyday screening and quantitation using Thermo Scientific TraceFinder software.
Confident identification for unknown small molecules
Confident decisions can only be made when using extensive, high-quality spectral libraries. With exhaustive and chemically diverse high-resolution MSn fragmentation spectra, know more unknowns using mzCloud, the world’s largest LC-MSn reference spectral library, delivering powerful characterization and identification capabilities.
Confident identification for unknown small molecules
For offline mzCloud access, or the ability to curate your own proprietary libraries, Thermo Scientific mzVault allows you to store and share knowledge across our qualitative and quantitative application packages to drive confident decision making.
Learn more
Go from spectra to structure and data to knowledge
No matter what your small molecule research application, from metabolomics to biomarker discovery, environmental and food safety, pharma metabolite or impurity identification, extractables and leachables to forensic or clinical toxicology, Compound Discoverer software offers a unique approach to small molecule structural identification analysis.
Learn more
Simplifying high-throughput screening and quantitation
Smart sample flagging, flexible data review, and custom report generation remove bottlenecks associated with data review and simplify sample analysis at each step of the process.
Learn more
Implement HRAM-MS within regulated environments with the necessary tools
Common applications for Orbitrap Exploris 120 Mass Spectrometer
To perform method development and everyday testing with confidence, you need a liquid chromatography mass spectrometry (LC-MS) workflow solution that is relevant to your laboratory’s goals. The Orbitrap Exploris 120 MS instrument can be used for a variety of applications, including clinical research, environmental and food safety, forensic toxicology, and pharmaceutical analysis.
Orbitrap Exploris 120 mass spectrometer ordering guide
Related products
Resources
Additional literature
Support
Additional resources
Our Products have not been tested or validated for such applications...
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.