TriPlus™ RSH Autosampler and Liquid Handling System
Actual product may vary
Thermo Scientific™
TriPlus™ RSH Autosampler and Liquid Handling System
Process a large number of samples quickly and reliably for your LC/MS analysis with the TriPlus RSH Autosampler and Liquid Handling System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
00950-01-00513
00950-01-00512
C0950-01-00610Autosampler (RTC Version)
Catalog number 00950-01-00513
Price (USD)
-
Maximize your productivity by processing large numbers of LC/MS samples quickly and reliably with the Thermo Scientific™ TriPlus™ RSH Autosampler and Liquid Handling System. This autosampler, which is equipped with Robotic Tool Change (RTC), takes liquid handling and sample preparation to the next level by automating switching between many different tools. The RTC enables unattended 24/7 operations, even for multistep workflows, by transferring repetitive time-consuming manual tasks to a robotic platform.
For liquid handling and sample preparation processes requiring different tools, the TriPlus RSH Autosampler and Liquid Handling System improves productivity and reduces sample handling time thanks to its automated and precision-driven RTC. The TriPlus RSH Autosampler and Liquid Handling System eliminates the manual labor and downtime traditionally required for time-consuming steps such as standard or stock sample dilution, or internal standard addition.
Other system benefits include:
Adaptability
  • Flexible capacity: 420 x 1 mL, 324 x 2 mL, or 90 x 10 mL vials, as well as other vial types and microtiter plates, can be loaded onto the temperature-controlled stack.
  • High throughput: Load up to 6912 samples from 18-microtiter plates with an optional six-drawer stack.(autosamplers with standard X-Axis only).
Reliability
  • Missing vials and correct positioning of the injector are tracked.
  • Monitoring of the temperature-controlled stack ensures that the drawer is in the correct position and the temperature is logged.
Flexibility
  • Meet any application need with simple liquid injection to complete workflows. Augment preparation capabilities by increasing sample capacity or adding modules.
User friendly
  • System seamlessly interfaces with Thermo Scientific™ Xcalibur™ or Thermo Scientific™ TraceFinder™  software so data acquisition starts quickly.
  • Use or optimize pre-developed methods with the optional TriPlus RSH Sampling Workflow Editor Software.
Specifications
DescriptionTriPlus RSH Autosampler with Robotic Tool Change for highest productivity and flexibility
Sample Capacity6x well-plates (96 and 384); 6x54 2 mL vials; 6x15 10 mL vials
Unit SizeEach