LC TriPlus™ RSH μSPE System for Sample Cleanup
LC TriPlus™ RSH μSPE System for Sample Cleanup
LC TriPlus™ RSH μSPE System for Sample Cleanup
LC TriPlus™ RSH μSPE System for Sample Cleanup
Thermo Scientific™
LC TriPlus™ RSH μSPE System for Sample Cleanup
Perform automated micro solid phase extraction fully integrated with LC-MS analysis.
Have Questions?
Catalog number C0950-01-00611
Price (USD)
-
Helping laboratories achieve reliable and high-throughput LC-MS analysis with the automated and online sample preparation capabilities of Thermo Scientific™ TriPlus™ RSH μSPE Autosampler. This system offers a fully automated online approach for sample clean-up while replacing time-consuming sample preparation steps. Suitable for a wide variety of matrices, including biological samples, the TriPlus μSPE system offers a unified method for LC-MS analysis. By using a single type of miniaturized cartridge, this system also offers the benefits of scaling down sample volume and solvent usage.
Improve laboratory productivity
  • Automation of time consuming sample preparation steps, and LC-MS integration
  • Reduced analytical variability
  • Minimized sample degradation
  • Limited sample size needed
  • Reduced solvent consumption – environmentally friendly
  • No concentration step needed
  • Accurate and precise LC-MS injections
  • Look-ahead functionality allowing for the simultaneous sample preparation and LC-MS run
Exceptional flexibility
TriPlus RSH μSPE systems are compatible with the following Thermo Scientific mass spectrometers:
  • Thermo Scientific™ TSQ™ series triple quadrupole mass spectrometers for the ultimate sensitivity, precision and accuracy in quantitative workflows
  • Thermo Scientific™ Q-Exactive™ MS,  Thermo Scientific™ Orbitrap Exploris™ MS and Thermo Scientific™ Orbitrap™ Tribrid series mass spectrometers combine full-scan MS/MS, superior resolution, and mass accuracy to ensure the highest possible confidence in every ID
Specifications
DescriptionLC TriPlus RSH μSPE autosampler
Injection Volume RangeUp to 100 μL
Temperature Range4°C - 40°C
Sample Capacity6x well-plates (96 and 384); 6x54 2 mL vials; 6x15 10 mL vials
Unit SizeEach