Orbitrap Tribrid Mass Spectrometers
Go beyond with Orbitrap Tribrid MS for large and small molecule analysis
Thermo Scientific Orbitrap Tribrid mass spectrometers enable high throughput analyses of challenging samples. Built on revolutionary Thermo Scientific Tribrid architecture, they combine the best of quadrupole, Orbitrap, and linear ion trap mass analyzers.
Go beyond with accurate and precise quantitation of low-abundance peptides in complex matrices, characterization of structural isomers, from drug metabolites to intact proteins, resolution of isobaric species, and native protein complexes structure characterization.
Which Orbitrap Tribrid MS is right for you?
Orbitrap Ascend Tribrid MS
Orbitrap Eclipse Tribrid MS
Orbitrap IQ-X Tribrid MS
Ultimate experimental throughput, versatility, and ease-of-use in multiomics, structural biology, and biopharma applications.
Ideally suited for proteomics, structural biology, small-molecule, and biopharmaceutical characterization experiments.
Reveal complex chemical structures for compound identification and structure elucidation of small molecules.
Applications
Multi-omics, TMT, intact/top-down proteomics, targeted quantification, structural biology, ligand binding, biotherapeutics, fusion proteins, and new modalities.
Multi-omics, TMT, intact top-down proteomics, targeted quantification, crosslinking, biotherapeutics.
Metabolomics, lipidomics, drug development Met ID, extractables and leachables, industrial environmental health and more.
Ease-of-use
  • Auto-ready calibration source
  • Scheduled remote calibration
  • One click button calibration
  • One click button calibration
  • Auto-ready calibration source
  • Scheduled remote calibration
  • One click button calibration
Orbitrap mass range (m/z)
40-6,000 (16,000 with Native MS)
40-6,000 (8,000 with HMR)
40-2,000
Advanced workflows
SQUAD, AcquireX, TMT and TMTpro (up to 32plex), TurboTMT, RTS/RTLS, APD, DIA-PTCR
SQUAD, AcquireX, TMT and TMTpro (up to 32plex), TurboTMT, RTS/RTLS, APD, DIA-PTCR
SQUAD, AcquireX, RTS/RTLS
Optional add-ons IC/ETD/PTCR/UVPD/Native MS
IC/ETD/PTCR/UVPD/HMR
IC/UVPD
Applications
Proteomics
Orbitrap Tribrid mass spectrometers are the ultimate tool for high accuracy quantitation and deep characterization. Explore individual proteoform-level functionality in complex biological systems.
Metabolomics and lipidomics
Get deeper analysis into the metabolome by combining discovery and targeted methods. Characterize small molecules and lipids with intelligent acquisitions & unique fragmentation options.
Structural biology
Determine biological functions with Orbitrap Tribrid MS, the complete package for characterization of protein-complexes. Handle experiments from peptides up to ligand binding with native complexes.
Biopharmaceuticals
Keep pace with biopharmaceutical advances, from fusion proteins to oligonucleotides to MetID. The diversity of needs lends to the versatility of the Orbitrap Tribrid MS.
System architecture that enables multiple applications
Orbitrap Tribrid mass spectrometer architecture enables experimental flexibility and high performance. With three mass analyzers, the Quadrupole, the Orbitrap, the duel linear ion traps, as well as the Orbitrap and ion trap, which provide two detectors, enable unique methodologies and parallelization within experiments.
The new standard in ease-of-use LCs for all flows
For researchers pursuing the next scientific breakthrough, Thermo Scientific Vanquish UHPLC systems deliver maximum performance 24/7 for reproducible nano-, capillary-, and/or micro-flow LC-MS. Combine the Vanquish Horizon, Duo, or Flex with your Biopharma applications, or use the new Vanquish Neo in nano or capillary flow for your multiomics or structural biology experiments.
We’ve got a column for that!
Solve your chromatography challenges with our extensive portfolio of HPLC and UHPLC columns. Bringing you a 40-year legacy of innovation, Thermo Scientific (U)HPLC columns offer an unmatched variety of chemistries, from C18 phases, to innovative mixed-mode chemistries, to the broadest range of solid core columns on the market, and more.
Ion mobility
The Thermo Scientific FAIMS Pro Duo interface extends differential ion mobility capability to multiple Thermo Scientific next-generation mass spectrometers, overcoming sample complexity and matrix interferences delivering increased productivity and data quality.
Connect projects, people, and instruments on a global level
Designed to support your digital transformation journey, the Thermo Scientific Ardia Platform is a scalable software solution for modern, dynamic (often cloud-based) environments. It connects and organizes projects, people, and instruments, on a global level, with web-based access and secure centralized data storage, enabling scientists to collaborate more efficiently and gain new insights faster, helping them get closer to the truth.
Services
Scalable service and support plan options
Discover flexible solutions to meet your specific service and support needs, from plans for single instruments to one that is customized to your entire lab or enterprise.
Resources
Orbitrap Ascend MultiOmics Tribrid Mass Spectrometer
Scale up your multiomics analyses with throughput, versatility and ease-of-use.
Orbitrap Ascend Structural Biology Tribrid Mass Spectrometer
Unlock the intricacies of complex molecular structures.
Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Set new standards in biotherapeutic characterization.
Orbitrap Eclipse Tribrid Mass Spectrometer
Go beyond today’s discoveries with power and versatility.
Orbitrap IQ-X Tribrid Mass Spectrometer
Designed for small-molecule analysis.
Orbitrap Ascend MS Year in Review
Scientific compendium covering multiomics, structural biology and biopharma applications.
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.