    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Isotype Controls ›
          • Rat IgG2b Isotype Controls
          Invitrogen
          Rat IgG2b Isotype Control, Pacific Orange™
          View all Rat IgG2b Isotype Controls
          Questions & Answers
          Cite Rat IgG2b Isotype Control, Pacific Orange™
          Additional Information:
          {{banner.description}}
          Product Details
          R2B30
          Applications
          Tested Dilution
          Flow Cytometry (Flow)
          Assay-Dependent
          Control (Ctrl)
          Assay-Dependent
          Product Specifications
          Host/Isotype
          Rat / IgG2b
          Class
          Monoclonal
          Type
          Isotype Control
          Conjugate
          Pacific Orange™ Pacific Orange™ Pacific Orange™
          Excitation/Emission Max
          400/551 nm View spectra spectra
          Form
          Liquid
          Concentration
          100 µg/mL
          Amount
          50 µg
          Purification
          purified
          Storage buffer
          PBS with 4mg/mL BSA
          Contains
          0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2556541
          Target Information
          Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.
          View product citations for antibody R2B30 on CiteAb
          Bioinformatics
          Protein Aliases: IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline