Oncomine cfNA Assays
Liquid Biopsy NGS Assays for Oncology Research
While tissue next-generation sequencing (NGS) is the standard for gaining insights into the biology of a tumor, liquid biopsy–based NGS is emerging as a highly sensitive method to understand resistance mechanisms as a tumor evolves. Liquid biopsies offer several advantages and can deliver a relevant genomic profile even if a tissue sample is unavailable, or complement tissue samples to understand tumor heterogeneity. Oncomine cell-free assays can help lead to important insights from liquid samples, including blood, bile, cerebrospinal fluid, and urine.
Key benefits inlcude:
  • A broad range of NGS assays for research applications across multiple solid tumor types
  • Easy, fast, and automated workflows to help minimize hands-on time and maximize efficiency
  • Integrated data analysis and reporting tools for easy data interpretation
Featured research solutions
Tissue and liquid biopsy solution: Oncomine Precision Assay on the Genexus System
The Oncomine Precision Assay on the Ion Torrent Genexus System is a focused multigene panel that detects a broad range of genomic alterations, including single nucleotide variants (SNVs), insertions and deletions (indels), copy number variations (CNVs), and fusions across 50 genes from solid tumor tissue and liquid biopsy samples.
Requiring only 20 minutes of hands-on time with two touchpoints, the highly automated system allow you to report NGS results in as little as 24 hours for integration with your immunohistochemistry results. In addition, a high success rate with low inputs means that more samples can be tested with results, even for small and challenging samples.
Figure 1. NGS workflow for the Oncomine Precision Assay on the Genexus System.
*Reporting solutions include Ion Torrent™ Oncomine Reporter, the laboratory’s own software, or other third-party software.
Oncomine Precision Assay customer webinars
On Demand Webinar: Oncomine Precision Assay: Automated and decentralized genomic profiling of plasma cell-free DNA
Hear Dr. Ting Chan, PhD from the University of Melbourne at OncomineWorld as she discusses the benefits of using liquid biopsy NGS for oncology research. 
Dr. Ting Chan, PhD
Curation Scientist, Centre for Cancer Research
University of Melbourne
Melbourne, Australia
On Demand Webinar: Analysis of central nervous system tumor ctDNA from cerebrospinal fluid
Dr. Leomar Ballester discusses using cerebrospinal fluid as an alternative to plasma samples for liquid biopsy NGS research.
Dr. Leomar Ballester, MD, PhD
Assistant Professor, Department of Pathology and Laboratory Medicine
MD Anderson Cancer Center
Liquid biopsy solution: Oncomine Pan-Cancer Cell-Free Assay on the Ion GeneStudio System
The Oncomine Pan-Cancer Cell-Free Assay on the Ion GeneStudio System is a focused multigene panel that detects a broad range of genomic alterations, including SNVs, indels, CNVs, and fusions across 52 genes from solid tumor liquid biopsy samples.
The highly automated, end-to-end NGS solution allows you to go from blood sample to report in just 2-3 days. In addition, multiple sample sources maybe be used such as bile, cerebro-spinal fluid, and urine.
Figure 2. NGS workflow for the Oncomine Pan-Cancer Cell-Free Assay on the GeneStudio S5 System.
*Reporting solutions include Ion Torrent™ Oncomine Reporter, the laboratory’s own software, or other third-party software.
The Oncomine cfNA assay portfolio
Multi-cancer and tumor-type specific gene coverage to fit your lab’s needs.
Oncomine
Precision
Assay
Oncomine Pan-Cancer Cell-Free Assay
Oncomine Lung Cell-Free Total Nucleic Acid Research Assay
Oncomine Breast cfDNA Research Assay v2
Oncomine
Colon cfDNA
Assay
Assay portfolio gene content
Multi-cancer
Oncomine Precision Assay
SNVs/Indels CNVs Fusions
AKT1 CDKN2A FGFR1 HRAS MTOR RAF1 ALK FGFR1 ALK FGFR3 RET
AKT2 CHEK2 FGFR2 IDH1 NRAS RET AR FGFR2 AR MET ROS1
AKT3 CTNNB1 FGFR3 IDH2 NTRK1 ROS1 CD274 FGFR3 BRAF NRG1 RSPO2
ALK EGFR FGFR4 KIT NTRK2 SMO CDKN2A KRAS EGFR NTRK1 RSPO3
AR ERBB2 FLT3 KRAS NTRK3 TP53 EGFR MET ESR1 NTRK2  
ARAF ERBB3 GNA11 MAP2K1 PDGFRA   ERBB2 PIK3CA FGFR1 NTRK3  
BRAF ERBB4 GNAQ MAP2K2 PIK3CA   ERBB3 PTEN FGFR2 NUTM1  
CDK4 ESR1 GNAS MET PTEN            
Oncomine Pan-Cancer Cell-Free Assay
SNVs/Indels CNVs Fusions
AKT1 CTNNB1 FGFR1 GNAS MAP2K2 PIK3CA SMO CCND1 FGFR1 ALK MET
ALK DDR2 FGFR2 HRAS MET PTEN TP53 CCND2 FGFR2 BRAF NTRK1
APC EGFR FGFR3 IDH1 MTOR RAF1   CCND3 FGFR3 ERG NTRK3
AR ERBB2 FGFR4 IDH2 NRAS RET   CDK4 MET ETV1 RET
ARAF ERBB3 FLT3 KIT NTRK1 ROS1   CDK6 MYC FGFR1 ROS1
BRAF ESR1 GNA11 KRAS NTRK3 SF3B1   EGFR   FGFR2  
CHEK2 FBXW7 GNAQ MAP2K1 PDGFRA SMAD4   ERBB2   FGFR3  
Lung Breast Colon
Oncomine cfTNA Assay Oncomine Breast cfDNA Assay v2 Oncomine Colon cfDNA Assay
ALK KRAS PIK3CA AKT1 ERBB3 KRAS AKT1 ERBB2 NRAS
BRAF MAP2K1 RET CCND1 ESR1 PIK3CA APC FBXW7 PIK3CA
EGFR MET ROS1 EGFR FBXW7 SF3B1 BRAF GNAS SMAD4
ERBB2 NRAS TP53 ERBB2 FGFR1 TP53 CTNNB1 KRAS TP53
            EGFR MAP2K1  
Experiences of Oncomine cfNA assay users
Dr. Brandon Sheffield, MD
Pathologist, W. Osler Health System, Brampton, Ontario
“The Oncomine Precision Assay, is novel…in that it also looks at circulating cell-free tumor derived RNA to detect fusions as opposed to most of the commercially available assays, which are looking only at DNA.”
Beatriz Bellosillo, PhD
Hospital del Mar, Spain
“Genomic heterogeneity after anti-EGFR therapy was successfully detected in 94% of the metastatic colorectal cancer research samples using a next-generation sequencing multi-biomarker approach. The Oncomine Colon cfDNA Assay enabled detection of somatic mutations with MAF >0.1%.
Ordering information
Oncomine cfNA Assays frequently asked questions
Oncomine cfNA Assays are targeted next-generation sequencing (NGS) research panels designed to sensitively detect genomic alterations in cell-free nucleic acids (cfNA), including cfDNA and cfRNA, from liquid biopsy samples (i.e, blood, bile, cerebrospinal fluid, and urine). The assays provide multi-biomarker insights including single nucleotide variants (SNVs), insertions/deletions (indels), copy number variations (CNVs), and fusions when tissue samples are not available, or as a complement to tissue testing to better understand tumor heterogeneity.
Oncomine cfNA Assays are highly sensitive, achieving a limit of detection (LOD) down to 0.1% frequency (with a sensitivity of >90% and specificity of >98%), or 1 mutant copy in a background of 1,000 wild-type (WT) copies. To achieve a 0.1% LOD, 20 ng of input cfDNA is recommended. Lower amounts of cfDNA can be used, but the limit of detection may be higher depending on the input amount.  
Oncomine cfNA Assays are used to detect mutations present in cfNA from plasma isolated from blood. cfNA is of interest because it harbors the same genomic alterations present in the tumor, such as SNVs, indels, CNVs, and fusions.  In addition, the liquid biopsy assays can be used repeatedly to assess for changes as the tumor evolves over time. While Oncomine cfNA Assays are generally used to evaluate blood samples, researchers may also choose to evaluate other sample types such as bile, cerebrospinal fluid, or urine. 
Oncomine cfNA Assays are designed for use with the Ion Torrent NGS platforms including the Genexus System and GeneStudio S5 Systems. The Genexus System is a fast, easy, automated two-instrument NGS platform to bring the power of rapid NGS to your lab without the need for specialized sequencing or bioinformatics expertise. The GeneStudio S5 System is a scalable, flexible, and high-throughput NGS platform for a broad range of applications to achieve insights fast and efficiently. 
Ready to speak to a Thermo Fisher Scientific representative?
We will be happy to answer your questions and provide a demo of our NGS solutions.
For Research Use Only. Not for use in diagnostic procedures.
PMR-003531