Epi5™ Episomal iPSC Reprogramming Kit
Invitrogen™
Epi5™ Episomal iPSC Reprogramming Kit
The Epi5™ Episomal iPSC Reprogramming Kit provides an easy-to-use, highly efficient set of 5 episomal vectors designed by Dr. OkitaRead more
Have Questions?
Catalog NumberQuantity
A159601 kit
Catalog number A15960
Price (USD)
1,875.65
Online Exclusive
1,934.00
Save 58.35 (3%)
Each
Quantity:
1 kit
Price (USD)
1,875.65
Online Exclusive
1,934.00
Save 58.35 (3%)
Each
The Epi5™ Episomal iPSC Reprogramming Kit provides an easy-to-use, highly efficient set of 5 episomal vectors designed by Dr. Okita in the laboratory of Professor Yamanaka at the Center for iPS Cell Research and Application (CiRA), Kyoto University. This system produces transgene-free, virus-free human induced pluripotent stem cells (iPSCs) with efficiencies in the range of 0.04% to 0.3%, depending upon the cell type being reprogrammed.
Features of the Epi5™ Episomal iPSC Reprogramming Kit include:
• Easy to use and highly efficient—no need for small molecules during reprogramming and the added p53 suppression provides enhanced iPSC generation (1)
• Usable for all stages of your iPSC research—transgene-free and viral-free reprogramming allows use from basic to pre-clinical research
• Flexibility in media systems—can be used in either feeder-free or feeder-based media systems
Create Transgene and Virus-free iPSCs
Episomal vectors are a well-described system for producing transgene-free, virus-free iPSCs, providing a source of iPSCs for all stages of your pluripotent stem cell research. Other reprogramming methods, such as lentivirus, contain transgenes that can integrate into the host genome, potentially disrupting the genome or causing unpredictable results.
As oriP/EBNA1 vectors, these episomal vectors contain 5 reprogramming factors (Oct4, Sox2, Lin28, Klf4, and L-Myc) and replicate extra-chromosomally only once per cell cycle. At this replication rate, the episomes are lost at a rate of approximately 5% per cell generation. This system shows enhanced iPSC generation through p53 suppression, and the inclusion of L-Myc has been shown to be more potent and specific then c-Myc during human iPSC generation (1).
Easy-to-Use System
This 2-vial kit provides the optimal system for episomal vector reprogramming and eliminates the need for use of small molecules in the reprogramming medium. The episomal vectors are introduced into the cell by electroporation. We recommend use of the Neon™Transfection System for electroporation, allowing flexibility in the volume and thus number of cells needed for reprogramming.
Flexibility in Selection of Media Systems
The Epi5™ Episomal iPSC Reprogramming Kit can be used with multiple cell types and media systems. Protocols for fibroblasts and CD34+ blood cells are available in either feeder or feeder-free based reprogramming.
1. Okita K, Matsumara Y, Sato Y, et al. A more efficient method to generate integration free human iPS cells. Nature Methods 8, 409-412, 2011.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodNone
Delivery TypeTransfection
For Use With (Application)Stem Cell Engineering & Reprogramming
For Use With (Equipment)Electroporator, Neon Transfection System
Key Functionsel
Product LineEpi5
Product TypeEpisomal iPSC Reprogramming Kit
PromoterCAG
Protein TagNone
Quantity1 kit
Replication OriginpUC Origin
Reporter GeneNone
Selection Agent (Eukaryotic)None
VectorEBNA OriP
Unit SizeEach
Contents & Storage
2 tubes. Store at -5 to -20°C.
Frequently asked questions (FAQs)
Can I use pen/strep or other antibiotics for episomal reprogramming of cells?
The use of antibiotics is not recommended during episomal reprogramming, especially Fungizone antimycotic. If antibiotics must be used during the reprogramming process, pen/strep could be used. For established iPSCs (after reprogramming is complete), either antibiotic/antimycotic or pen/strep should be fine.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I reprogram mouse cells with the Epi5 Episomal iPSC Reprograming Kit (Cat. No. A15960)?
No. The Epi5 Episomal iPSC Reprogramming Kit (Cat. No. A15960) was designed to reprogram human cells, not mouse cells. We currently only have data with this kit on reprogramming human cells, which aligns with the use of EBNA/oriP episomal vectors mainly in primate cells. Although rodent cells lack the oriP replicon and are not permissive, some studies replaced or modified oriP with other sequences to mediate replication and some literature reported the use of unmodified EBNA/oriP in certain hamster and mouse lines.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When generating iPSCs using the Epi5 Episomal iPSC Reprogramming Kit (Cat. No. A15960), how confluent should my cells be on the day of transfection?
When generating iPSCs using the Epi5 Episomal iPSC Reprogramming Kit (Cat. No. A15960) cells should be approximately 75-90% confluent on the day of transfection (Day 0). Since over-confluency results in decreased transfection efficiency, we recommend replating overly confluent cells to achieve 75-90% confluency on the day of transfection.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How many reprogramming experiments can I perform with the Epi5 Episomal iPSC Reprogramming Kit (Cat. No. A15960)?
The Epi5 Episomal iPSC Reprogramming Kit (Cat. No. A15960) provides enough material to process 20 reprogramming experiments (each reprogramming experiment requires 1 × 105 cells and 1 µL each of the Epi5 Reprogramming Vectors and the Epi5 p53 & EBNA Vectors). Each vial of vectors in the kit contains 20 µL, and you will use 1 µL (from each vial) per experiment. Before use, briefly centrifuge the thawed vectors to collect them at the bottom of the tube.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which kits do you offer for stem cell reprogramming?
We offer the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit (Cat. Nos. A16517, A16518) which is a non-integrating system that uses Sendai virus vectors to reprogram somatic cells into induced pluripotent stem cells (iPSCs). The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit contains three Invitrogen CytoTune 2.0 reprogramming vectors that are used for delivering and expressing key genetic factors necessary for reprogramming somatic cells into iPSCs. Only one application of the vectors is required for successful reprogramming.
Additionally, we offer the Episomal iPSC Reprogramming Vectors (Cat. No A14703) or Epi5 Episomal iPSC Reprogramming Kit (Cat. No A15960). This is a non-integrating system that reprograms somatic cells into induced pluripotent stem cells (iPSCs). These two products contain a mixture of six or five vectors designed to provide the optimal system for generating transgene-free and virus-free iPSCs in a feeder-free environment. Originally developed by Junying Yu and James Thomson (http://www.ncbi.nlm.nih.gov/pubmed/19325077) and further optimized by Cellular Dynamics International, these Episomal iPSC Reprogramming Vectors have proven successful in reprogramming a number of different somatic cell types.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Citations & References (2)
Citations & References
Abstract
An optogenetic approach for assessing formation of neuronal connections in a co-culture system.
Authors:Su CT, Yoon SI, Marcy G, Chin EW, Augustine GJ, Goh EL,
Journal:
PubMed ID:25742527
'Here we describe a protocol to generate a co-culture consisting of 2 different neuronal populations. Induced pluripotent stem cells (iPSCs) are reprogrammed from human fibroblasts using episomal vectors. Colonies of iPSCs can be observed 30 days after initiation of fibroblast reprogramming. Pluripotent colonies are manually picked and grown in neural ... More
Generation of human iPSCs from cells of fibroblastic and epithelial origin by means of the oriP/EBNA-1 episomal reprogramming system.
Authors:Drozd AM, Walczak MP, Piaskowski S, Stoczynska-Fidelus E, Rieske P, Grzela DP,
Journal:
PubMed ID:26088261
The prospect of therapeutic applications of the induced pluripotent stem cells (iPSCs) is based on their ability to generate virtually any cell type present in human body. Generation of iPSCs from somatic cells has opened up new possibilities to investigate stem cell biology, to better understand pathophysiology of human diseases, ... More