HAAKE™ MARS™ iQ Rotational Rheometer
HAAKE™ MARS™ iQ Rotational Rheometer
HAAKE™ MARS™ iQ Rotational Rheometer
HAAKE™ MARS™ iQ Rotational Rheometer
HAAKE™ MARS™ iQ Rotational Rheometer
Thermo Scientific™
HAAKE™ MARS™ iQ Rotational Rheometer
Thermo Scientific™ HAAKE™ MARS™ iQ Rotational Rheometers ensure intuitive, reliable and flexible rheological measurements. Everything about this QC rheometer is designed to provide intuitive handling for reliable and fast results. Discover more iQ for your quality control!
Have Questions?
Change viewbuttonViewtableView
Catalog NumberRotational Speed Range
379-07000.001 to 2000 rpm
379-08000.01 rpm to 2000 rpm
Catalog number 379-0700
Price (USD)
-
Rotational Speed Range:
0.001 to 2000 rpm
Thermo Scientific™ HAAKE™ MARS™ iQ Rotational Rheometers ensure intuitive, reliable and flexible rheological measurements. Everything about this QC rheometer is designed to provide intuitive handling and fast, reliable results. A sophisticated touchscreen enables standard operating procedures to be run right from rheometer itself. Connect Assist functionality guides users for failure-free measurement.
Choose a HAAKE MARS iQ Rheometer with either the ball bearing or air bearing system to suit your lab. A broad range of accessories helps meet the ever-greater testing demands on QC labs.
HAAKE MARS iQ Rotational Rheometers are the replacements for the discontinued Thermo Scientific™ HAAKE™ RheoStress™ 1 Rheometer and Thermo Scientific™ HAAKE™ RotoVisco™ 1 Rotational Rheometer.
HAAKE MARS iQ Rheometers offer many advantages:
Intuitive Operations
  • Multifunctional 7' touchscreen for instrument operation and SOP execution right at your fingertips
  • 'Assist' functionalities to simplify operation and prevent mistakes
  • 'Connect Assist' quick coupling of measuring geometries and temperature modules with automatic recognition
  • 'Color Assist' color-coded plugs for quick, correct selection of temperature modules
  • HAAKE™ RheoWin™ Rheology Software for extensive measuring flexibility with free updates (Optional 21 CFR part 11 module available)
Intelligent Design
  • Folded H frame concept for greater stiffness and ultra-precise lift control with a wide travel range
  • Durable material: high-performance mineral composite casting with high vibrational damping, minimal temperature expansion and high chemical resistance
  • Next generation of highly dynamic, powerful EC motor with a mechanical or air bearing.
Individualized Configurations
  • Broad scope of temperature modules, measuring geometries and application-oriented measuring cells, e.g., pressure cells, building material measuring cell or tribology measuring cell
  • Sensitive normal force sensor measures axial forces in both directions for applications such as texture analysis
  • Lateral and bottom access for customized test requirements
Application solutions
The HAAKE MARS iQ Rheometers are well-suited for the rheological characterization of a broad range of samples from low-viscous or water-like up to stiff and paste-like semisolids:
  • Investigation of paints, coatings and inks: shear viscosity, thixotropy, yield stress, shear recovery, levelling and sagging behavior
  • Rheology and textural properties of foods and foodstuffs: shear viscosity, yield stress, tribology, texture analysis (axial bending, breaking, squeezing tests), phase transition
  • Rheology of cosmetics and pharmaceuticals: viscosity, yield stress, viscoelastic properties, stability tests
  • Rheology of polymer (e.g. polymer melts and solutions): viscoelasticity, phase transitions (glass transition, crystallization, melting), processing behavior (hot melt extrusion, molding, spinning), molecular weight, curing
  • Rheology of petrochemicals (crude oils, fracking liquids and drilling fluids): viscosity, yield stress, pressure dependent properties, stability tests, wax deposition, wax appearance temperature (WAT)
  • Dynamic shear rheometer (DSR) tests according to AASHTO, ASTM and DIN used for bitumen and asphalt
  • Construction and building material (cement paste and slurries): flow properties of suspensions with large particles
Specifications
Angular Resolution0.63 μrad
DescriptionHAAKE MARS iQ Rheometer - air-bearing
Dimensions (L x W x H)480 x 390 x 670 mm
Frequency Range0.0001 to 100 Hz
Normal Force(N): 0.01 to 50
Normal force resolution (N): 0.001
Rotational Speed Range0.001 to 2000 rpm
Temperature Range (Metric)-60°C to +400°C
Torque RangeMin. torque rotation: 0.001 mNm
Max. torque rotation: 150 mNm
Min. torque oscillation: 0.001 mNm
Max torque oscillation: 150 mNm
Torque resolution: 0.007 μNm
Velocity Range0.0001 to 209.4 rad/s
Weight (Metric)57 kg
Unit SizeEach